Skip to Content
Merck
All Photos(4)

Documents

WH0000142M1

Sigma-Aldrich

Monoclonal Anti-PARP1 antibody produced in mouse

clone 3G4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

PARP1 Antibody - Monoclonal Anti-PARP1 antibody produced in mouse, Parp1 Antibody, Anti-ADPRT, Anti-ADPRT1, Anti-PARP, Anti-PARP1, Anti-PPOL, Anti-pADPRT1, Anti-poly (ADP-ribose) polymerase family, member 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARP1(142)

General description

Poly (ADP-ribose) polymerase 1 (PARP1) is a nuclear protein and belongs to the PARP family. This protein is made up of the N-terminal DNA-binding domain, central auto modification domain and C-terminal catalytic domain. PARP1 protein is located on the nucleoli. The PARP1 gene is located on the human chromosome at 1q42.12.

Immunogen

PARP1 (AAH37545, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAESSDKLYRVEYAKSGRASCKKCSESIPKDSLRMAIMVQSPMFDGKVPHWYHFSCFWKVGHSIRHPDVEVDGFSELRWDDQQKVKKTAEAGGVTGKGQD

Application

Monoclonal Anti-PARP1 antibody produced in mouse has been used in:
  • western blotting
  • indirect immunofluorescence
  • high-throughput cellular thermal shift assay (CESTA HT)

Biochem/physiol Actions

Poly (ADP-ribose) polymerase 1 (PARP1) protein plays a role in DNA repair by catalyzing the polymerization of adenosine diphosphate (ADP)-ribose units. This protein also plays a role in the early response to DNA damage. Mutations in the PARP1 gene is associated with loss of cell viability.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingbiao Ji et al.
Current opinion in genetics & development, 20(5), 512-518 (2010-07-02)
Cell growth and differentiation during developmental processes require the activation of many inducible genes. However, eukaryotic chromatin, which consists of DNA and histones, becomes a natural barrier impeding access to the functional transcription machinery. To break through the chromatin barrier
Esin Orhan et al.
Cancer letters, 589, 216820-216820 (2024-04-05)
One in three Triple Negative Breast Cancer (TNBC) is Homologous Recombination Deficient (HRD) and susceptible to respond to PARP inhibitor (PARPi), however, resistance resulting from functional HR restoration is frequent. Thus, pharmacologic approaches that induce HRD are of interest. We
Arnab Ray Chaudhuri et al.
Nature reviews. Molecular cell biology, 18(10), 610-621 (2017-07-06)
Cells are exposed to various endogenous and exogenous insults that induce DNA damage, which, if unrepaired, impairs genome integrity and leads to the development of various diseases, including cancer. Recent evidence has implicated poly(ADP-ribose) polymerase 1 (PARP1) in various DNA
Michèle Rouleau et al.
Nature reviews. Cancer, 10(4), 293-301 (2010-03-05)
Recent findings have thrust poly(ADP-ribose) polymerases (PARPs) into the limelight as potential chemotherapeutic targets. To provide a framework for understanding these recent observations, we review what is known about the structures and functions of the family of PARP enzymes, and
Y Liu et al.
Cancer gene therapy, 24(5), 208-214 (2017-03-11)
Cisplatin resistance hinders the efficacy of chemotherapy in ovarian cancer. MicroRNAs (miRs) have been implicated in drug resistance in anti-cancer chemotherapy. We compared the expression profiles of miRs between cisplatin-resistant and cisplatin-sensitive ovarian cancer cells, and found that miR-216b was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service