Skip to Content
Merck
All Photos(1)

Key Documents

T6140

Sigma-Aldrich

N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt

plasmin substrate, chromogenic, ≥98% (TLC), powder

Synonym(s):

N-Tosylglycyl-L-prolyl-L-lysine 4-nitroanilide acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C26H34N6O7S · C2H4O2
CAS Number:
Molecular Weight:
634.70
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt, plasmin substrate

Assay

≥98% (TLC)

form

powder

solubility

ethanol: 50 mg/mL, colorless to yellow

storage temp.

−20°C

SMILES string

CC(O)=O.Cc1ccc(cc1)S(=O)(=O)NCC(=O)N2CCC[C@H]2C(=O)N[C@@H](CCCCN)C(=O)Nc3ccc(cc3)[N+]([O-])=O

InChI

1S/C26H34N6O7S.C2H4O2/c1-18-7-13-21(14-8-18)40(38,39)28-17-24(33)31-16-4-6-23(31)26(35)30-22(5-2-3-15-27)25(34)29-19-9-11-20(12-10-19)32(36)37;1-2(3)4/h7-14,22-23,28H,2-6,15-17,27H2,1H3,(H,29,34)(H,30,35);1H3,(H,3,4)/t22-,23-;/m0./s1

InChI key

KPGSNEYSJPIZCW-SJEIDVEUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt has been used:
  • as a substrate for examining the amidolytic activity of aprE2, aprE5-41
  • as a substrate for serine protease and fibrinogenolytic assays
  • as a substrate for plasmin assay

Biochem/physiol Actions

N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt is a chromogenic substrate for plasmin assay, serine protease enzyme activity and fibrinogenolytic assays.

Substrates

A chromogenic substrate for plasmin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Wang et al.
International journal of biological macromolecules, 146, 781-789 (2019-11-16)
The heterologous nature of SAK, a thrombolytic drug, elicits high titers of neutralizing antibodies, which limits its clinical use. Here, we aim to establish a SAK mutant with equivalent activity to the wild type but reduced antigenicity, which may allow
Overexpression of aprE2, a Fibrinolytic Enzyme Gene from Bacillus subtilis CH3-5, in Escherichia coli and the Properties of AprE2.
Jeong SJ, et al.
Journal of microbiology and biotechnology, 24(7), 969-978 (2014)
Streptococcus pneumoniae choline-binding protein E interaction with plasminogen/plasmin stimulates migration across the extracellular matrix.
Attali C, et al.
Infection and Immunity, 76(2), 466-476 (2008)
Michał Śmiga et al.
BMC microbiology, 19(1), 127-127 (2019-06-13)
Porphyromonas gingivalis is considered a keystone pathogen responsible for chronic periodontitis. Although several virulence factors produced by this bacterium are quite well characterized, very little is known about regulatory mechanisms that allow different strains of P. gingivalis to efficiently survive
Purification and characterization of a major fibrinolytic enzyme from Bacillus amyloliquefaciens MJ5-41 isolated from Meju
Jo HD, et al.
Journal of microbiology and biotechnology, 21(11), 1166-1173 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service