Skip to Content
Merck
All Photos(1)

Key Documents

U7129

Sigma-Aldrich

Urea solution

40 % (w/v) in H2O

Synonym(s):

Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

sterility

non-sterile; sterile-filtered

Quality Level

contains

0.1% benzoic acid as preservative

packaging

vial of 5 mL

concentration

40 % (w/v) in H2O

storage temp.

2-8°C

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Recommended for the detection of urease activity.

Biochem/physiol Actions

Urea solution is primarily used for protein denaturation. It also increases solubility of hydrocarbons and reduce micelle formation.[1] Urea solution at high concentration leads to the destabilization of amyloid β16−22 oligomers.

Preparation Note

Warm the refrigerated 40% urea solution to room temperature. Aseptically add 5 mL to 95 mL of sterile molten Urea Agar Base (Christensen). Mix well and dispense in sterile tubes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aqueous urea solutions: structure, energetics, and urea aggregation
Stumpe MC, et al.
The Journal of Physical Chemistry B, 111(22), 6220-6228 (2007)
L Li et al.
Cell death and differentiation, 22(7), 1081-1093 (2014-12-20)
P53 is critically important in preventing oncogenesis but its role in inflammation in general and in the function of inflammatory macrophages in particular is not clear. Here, we show that bone marrow-derived macrophages exhibit endogenous p53 activity, which is increased
G L French et al.
Lancet (London, England), 2(8551), 117-119 (1987-07-18)
Tuberculostearic acid, a structural component of Mycobacterium tuberculosis, was identified by gas chromatography/mass spectrometry with selected ion monitoring in cerebrospinal fluid (CSF) from 13 patients with proven and 8 out of 9 patients with suspected tuberculous meningitis; the negative result
A mechanically and electrically self-healing supercapacitor.
Hua Wang et al.
Advanced materials (Deerfield Beach, Fla.), 26(22), 3638-3643 (2014-02-21)
Sreenivasa C Ramaiahgari et al.
Archives of toxicology, 88(5), 1083-1095 (2014-03-07)
Immortalized hepatocyte cell lines show only a weak resemblance to primary hepatocytes in terms of gene expression and function, limiting their value in predicting drug-induced liver injury (DILI). Furthermore, primary hepatocytes cultured on two-dimensional tissue culture plastic surfaces rapidly dedifferentiate

Questions

  1. Hallo, wieviel ml sind in dem VIAL?

    1 answer
    1. As mentioned in the 'PROPERTIES' section of the Product Detail Page, this product consists of 5 mL per vial.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service