Skip to Content
Merck
All Photos(1)

Key Documents

51463

Millipore

Urea Broth

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74
Pricing and availability is not currently available.

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

dipotassium hydrogen phosphate, 9.5 g/L
phenol red, 0.01 g/L
potassium dihydrogen phosphate, 9.1 g/L
urea, 20 g/L
yeast extract, 0.1 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbe id | specific enzyme detection: suitable
microbiological culture: suitable

final pH

6.8±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

nonselective and identification for Enterobacter spp.
nonselective and identification for Escherichia coli
nonselective and identification for Klebsiella spp.
nonselective and identification for Proteus spp.
nonselective and identification for Salmonella spp.
nonselective and identification for coliforms
nonselective and identification for enterobacteriaceae

Application

Differential medium for the detection of urea-utilizing microorganisms, acc. to Rustigian and Stuart (1941).

Preparation Note

Dissolve 38.7 g in 1 litre distilled water and filter sterilize. This medium must NOT be heated.

Other Notes

Adaption of urea broth for radiosterilization[1][2][3]

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Rustigian et al.
Proceedings of the Society For Experimental Biology and Medicine. Society For Experimental Biology and Medicine, 47, 108-108 (1941)
E Eisenberg et al.
The Journal of applied bacteriology, 58(1), 21-25 (1985-01-01)
Urea broth, a medium used for the identification of the genus Proteus, was sterilized by gamma radiation, using radiation doses of 1-1.5 Mrad. The radiation-sterilized medium, modified by adding sodium ascorbate and increasing its phenol red and yeast extract content
W.W. Feguson et al.
The Journal of Laboratory and Clinical Medicine, 28, 1715-1715 (1943)

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service