Skip to Content
Merck
All Photos(1)

Key Documents

U6883

Sigma-Aldrich

cis-Urocanic acid

≥98% (HPLC), solid (fluffy)

Synonym(s):

cis-3-(1H-imidazol-4-yl)-2-propenoic acid, cis-UCA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Product Name

cis-Urocanic acid, ≥98% (HPLC), solid (fluffy)

Quality Level

Assay

≥98% (HPLC)

form

solid (fluffy)

storage condition

protect from light

color

white

solubility

H2O: >10 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)\C=C/c1c[nH]cn1

InChI

1S/C6H6N2O2/c9-6(10)2-1-5-3-7-4-8-5/h1-4H,(H,7,8)(H,9,10)/b2-1-

InChI key

LOIYMIARKYCTBW-UPHRSURJSA-N

Application

cis-Urocanic acid (cis-UCA) has been used:
  • to study its protective effects on autoimmune encephalomyelitis
  • in sample preparation to study the distribution of skin chemical components by line illumination Raman microscopy technique in Line-focus illumination mode
  • as a standard to study the levels of cis-UCA and to check its effect on immune response in multiple sclerosis patients

Biochem/physiol Actions

cis-Urocanic acid (cis-UCA) affects the immune cells of the body and is excreted in the urine due to photoisomerization. It inhibits tumor immunity through photocarcinogenesis in mice. cis-Urocanic acid has been studied to prevent UV-radiation-induced DNA damage leading to skin cancer.
cis-Urocanic acid, a 5-HT agonist, is a product of UV isomerization of trans-urocanic acid in skin, which leads to immunosuppression via activation of 5-HT2A receptor.

Preparation Note

cis-Urocanic acid is soluble in water at a concentration that is greater than 10 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

High-speed line-focus Raman microscopy with spectral decomposition of mouse skin
Ilchenko O, et al.
Vibrational Spectroscopy, 83, 180-190 (2016)
S Beissert et al.
Journal of immunology (Baltimore, Md. : 1950), 167(11), 6232-6238 (2001-11-21)
UV radiation induces skin cancer primarily by its DNA-damaging properties, but also by its capacity to suppress the immune system. The photoisomer of urocanic acid (UCA), cis-UCA, is an important mediator of UV-induced immunosuppression and is involved in the inhibition
Jorge Correale et al.
Journal of neuroimmunology, 261(1-2), 134-140 (2013-06-27)
The role of cis-urocanic acid (UCA) as a UV-mediated immunomodulator in MS patients was investigated. Plasma levels of cis-UCA were significantly lower in MS patients compared to controls. Stimulation of MBP- and MOG-specific T cells in the presence of cis-UCA
Amy A Irving et al.
BMC neuroscience, 18(1), 8-8 (2017-01-07)
Ultraviolet B irradiation confers strong resistance against experimental autoimmune encephalomyelitis, a model of multiple sclerosis. This protection by ultraviolet B is independent of vitamin D production but causes isomerization of urocanic acid, a naturally occurring immunosuppressant. To determine whether UCA
Evgeny Berdyshev et al.
The Journal of allergy and clinical immunology, 147(1), 361-367 (2020-07-03)
The nonlesional skin of children with atopic dermatitis (AD) with peanut allergy (PA) is associated with increased transepidermal water loss; low urocanic acid (UCA) and pyrrolidone carboxylic acid (PCA), both of which are filaggrin breakdown products; and a reduced ratio

Articles

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service