Skip to Content
Merck
All Photos(1)

Key Documents

77951

Sigma-Aldrich

Imidazolepropionic acid

≥98.0% (GC)

Synonym(s):

1H-Imidazole-4-propanoic acid, 1H-Imidazole-5-propanoic acid, 3-(1H-Imidazol-4-yl)propionic acid, 3-(Imidazol-4(5)-yl)propionic acid, Deamino-histidine, Dihydrourocanic acid, NSC 66737

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (GC)

form

powder

storage temp.

2-8°C

SMILES string

OC(CCC1=CN=CN1)=O

InChI

1S/C6H8N2O2/c9-6(10)2-1-5-3-7-4-8-5/h3-4H,1-2H2,(H,7,8)(H,9,10)

InChI key

ZCKYOWGFRHAZIQ-UHFFFAOYSA-N

Biochem/physiol Actions

Imidazolepropionic acid is a product of histidine metabolism, which may involve oxidation or transamination

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A nitrogen-15 nuclear magnetic resonance study of the acid-base and tautomeric equilibriums of 4-substituted imidazoles and its relevance to the catalytic mechanism of .alpha.-lytic protease.
Roberts, J. D. , et al.
Journal of the American Chemical Society, 104, 3945-3949 (1982)
A covalent nicotinamide adenine dinucleotide intermediate in the urocanase reaction.
L H Matherly et al.
Biochemistry, 21(11), 2789-2794 (1982-05-25)
I Antener et al.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition, 53(2), 199-209 (1983-01-01)
The present study, as a part of a broader investigation on protein-energy-malnutrition (PEM) in rural Zaire, was undertaken in order to clarify varying aspects of histidine metabolism in patients suffering from protein-energy malnutrition (PEM). Measurement of histidine and its derivatives
Imidazolepropionic acid as a urinary metabolite of L-histidine.
N P SEN et al.
Biochemical and biophysical research communications, 9, 257-261 (1962-10-17)
Histidine metabolism in the human adult: histidine blood tolerance, and the effect of continued free L-histidine ingestion on the concentration of imidazole compounds in blood and urine.
W D Block et al.
The Journal of nutrition, 91(2), 189-194 (1967-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service