Skip to Content
Merck
All Photos(1)

Key Documents

U6508

Sigma-Aldrich

UCN-01

≥97% (HPLC), powder

Synonym(s):

KW-2401

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H26N4O4
CAS Number:
Molecular Weight:
482.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

optical activity

[α]20/D 130 to 160°, c = 0.2 in DMSO

storage condition

protect from light

color

light yellow

solubility

DMSO: >5 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

[H][C@@]1(C[C@@H](NC)[C@H]2OC)N3C4=C(N([C@@]2(C)O1)C5=C6C=CC=C5)C6=C7C(C(N[C@@H]7O)=O)=C4C8=C3C=CC=C8

InChI

1S/C28H26N4O4/c1-28-25(35-3)15(29-2)12-18(36-28)31-16-10-6-4-8-13(16)19-21-22(27(34)30-26(21)33)20-14-9-5-7-11-17(14)32(28)24(20)23(19)31/h4-11,15,18,25,27,29,34H,12H2,1-3H3,(H,30,33)/t15-,18-,25-,27-,28+/m1/s1

InChI key

PBCZSGKMGDDXIJ-HQCWYSJUSA-N

Related Categories

Application

UCN-01 has been used to study checkpoint kinase-1 (CHK1) protein function in trophoblast stem (TS) cells. It has also been used to study UV-induced CHK1 phosphorylation in IB10 cells.

Biochem/physiol Actions

UCN-01 is a selective inhibitor for conventional PKC isotypes over novel PKC isotypes.

Features and Benefits

This compound is featured on the PKB/Akt and PKC pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

UCN-01 is soluble in DMSO at a concentration that is greater than 5 mg/ml. However, it is insoluble in water.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sung-Bau Lee et al.
Science advances, 4(8), eaat4985-eaat4985 (2018-08-14)
DNA sequence and epigenetic information embedded in chromatin must be faithfully duplicated and transmitted to daughter cells during cell division. However, how chromatin assembly and DNA replication are integrated remains unclear. We examined the contribution of the Tousled-like kinases 1
Wuli Zhao et al.
Journal of experimental & clinical cancer research : CR, 37(1), 98-98 (2018-05-08)
A35 is a novel synthetic cyclizing-berberine recently patented as an antitumor compound. Based on its dual targeting topoisomerase (top) activity, A35 might overcome the resistance of single-target top inhibitors and has no cardiac toxicity for not targeting top2β. In this
Motofumi Suzuki et al.
Translational oncology, 10(4), 491-500 (2017-05-28)
Checkpoint kinase 1 (Chk1) is an evolutionarily conserved serine/threonine kinase that plays an important role in G
Bo Gao et al.
Carcinogenesis, 41(8), 1123-1133 (2019-11-20)
Isochorismatase domain-containing 1 (ISOC1) is a coding gene that contains an isochorismatase domain. The precise functions of ISOC1 in humans have not been clarified; however, studies have speculated that it may be involved in unknown metabolic pathways. Currently, it is
S Akinaga et al.
Anti-cancer drug design, 15(1), 43-52 (2000-07-11)
UCN-01 (7-hydroxystaurosporine) is a protein kinase inhibitor which is under development as an anti-cancer agent in the USA and Japan. Although UCN-01 was originally isolated from the culture broth of Streptomyces sp. as a protein kinase C-selective inhibitor, its ultimate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service