200499
Bexarotene
A retinoid X receptor (RXR) agonist that can permeate the blood-brain barrier and rapidly reduce Aβ40 and Aβ42 levels in APP/PS1 mice in an ApoE-dependent manner.
Synonym(s):
Bexarotene, 4-(1-(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)vinyl)benzoic acid
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white to pale yellow
solubility
DMSO: 50 mg/mL, clear, colorless
shipped in
ambient
storage temp.
−20°C
InChI
1S/C24H28O2/c1-15-13-20-21(24(5,6)12-11-23(20,3)4)14-19(15)16(2)17-7-9-18(10-8-17)22(25)26/h7-10,13-14H,2,11-12H2,1,3-6H3,(H,25,26)
InChI key
NAVMQTYZDKMPEU-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service