Skip to Content
Merck
All Photos(4)

Key Documents

T5251

Sigma-Aldrich

D-(+)-Trehalose dihydrate

≥98.5% (HPLC), from Saccharomyces cerevisiae

Synonym(s):

α,α-Trehalose dihydrate, D-Trehalose dihydrate, α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33
Beilstein:
5322018
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

Saccharomyces cerevisiae

Quality Level

Assay

≥98.5% (HPLC)

form

powder

optical activity

[α]/D 174 to 182 °, c = 1.0% (w/v) in water

technique(s)

HPLC: suitable
cryopreservation: suitable

mp

97-99  °C

application(s)

agriculture
agriculture
cell analysis
genomic analysis
life science and biopharma

SMILES string

[H]O[H].[H]O[H].OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11.2H2O/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12;;/h3-20H,1-2H2;2*1H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-;;/m1../s1

InChI key

DPVHGFAJLZWDOC-PVXXTIHASA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Trehalose, a disaccharide, acts as a chemical chaperone and serves as a carbohydrate reserve in microorganisms. It functions as an osmolyte, safeguarding cells against various environmental stresses. Further, it also plays a crucial role in protein stabilization during freeze drying, making it a valuable asset in the areas of biochemical, cell culture, and excipient research

Application

  • Use as a cryoprotectant in a variety of cell freezing media
  • Used in excipeint research

Biochem/physiol Actions

Trehalose is a non-reducing sugar formed from two glucose units joined by a 1-1 alpha bond. It is thought to provide plants and animals with the ability to withstanding periods of dehydration.

Features and Benefits

  • Prepared from Saccharomyces cerevisiae
  • Suitable for use with HPLC and GC-MS(4)

Preparation Note

Prepared by the modified procedure of Payen, R., Can. J. Res., 27B, 749 (1949).

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of longitudinal dynamics of trehalose?water solutions by inelastic ultraviolet scattering
A. Cesaro et al.
Philosophical Magazine, 87, 623-630 (2007)
Trehalose is a potent PCR enhancer: lowering of DNA melting temperature and thermal stabilization of taq polymerase by the disaccharide trehalose
Andrej-Nikolai S
Clinical Chemistry, 50, 1256-1259 (2004)
Dextran-Based Oxygen Nanobubbles for Treating Inner Retinal Hypoxia
Muhammad F
ACS Applied Nano Materials, 4, 6583?6593-6583?6593 (2021)
Sandra Jesus et al.
AAPS PharmSciTech, 19(1), 101-113 (2017-06-15)
This report extensively explores the benefits of including chitosan into poly-ε-caprolactone (PCL) nanoparticles (NPs) to obtain an improved protein/antigen delivery system. Blend NPs (PCL/chitosan NPs) showed improved protein adsorption efficacy (84%) in low shear stress and aqueous environment, suggesting that
Dami Kim et al.
The Analyst, 145(16), 5491-5499 (2020-07-01)
This study describes a microfluidic paper-based analytical device (μPAD) for separating plasma from whole blood and measuring glucose concentration. A two-dimensional μPAD was fabricated by wax printing, using chromatographic paper and was functionalized by the incorporation of chitosan into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service