Skip to Content
Merck
All Photos(3)

Documents

T4257

Sigma-Aldrich

Tripentadecanoin

≥99%

Synonym(s):

TG(15:0/15:0/15:0), 1,2,3-Tripentadecanoylglycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H92O6
CAS Number:
Molecular Weight:
765.24
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCC

InChI

1S/C48H92O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-46(49)52-43-45(54-48(51)42-39-36-33-30-27-24-21-18-15-12-9-6-3)44-53-47(50)41-38-35-32-29-26-23-20-17-14-11-8-5-2/h45H,4-44H2,1-3H3

InChI key

CLJLWABDLPQTHL-UHFFFAOYSA-N

Application


  • A rare natural lipid induces neuroglobin expression to prevent amyloid oligomers toxicity and retinal neurodegeneration.: This study explores the protective effects of a rare natural lipid, presumably including Tripentadecanoin, on neurodegeneration. It specifically focuses on neuroglobin expression to combat amyloid oligomers′ toxicity in the retina (Oamen et al., 2022).

  • Analysis of fatty acid content and composition in microalgae.: This research details methods for analyzing fatty acid content and composition in microalgae, providing insights into how Tripentadecanoin and similar fatty acids are profiled in these organisms (Breuer et al., 2013).


Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antonio Gázquez et al.
The Journal of physiology, 597(19), 4959-4973 (2019-07-10)
Placental structure and function can be modified as a result of maternal obesity affecting materno-fetal fatty acids (FA) transport. We report for the first time, in humans and in vivo, the kinetics of placental FA transfer in normo-weight and in
Alexandra D George et al.
Scientific reports, 10(1), 9255-9255 (2020-06-11)
Human milk provides the infant with the essential nutritive and non-nutritive factors required for health, growth and development. The human milk lipidome is complex, but comprises predominantly triacylglycerides. Historically, the fatty acid profile of the entire human milk lipidome has
R Whiteford et al.
Journal of applied microbiology, 131(3), 1305-1317 (2021-01-24)
The effects of combinatorial stress on lipid production in Phanerochaete chrysosporium remain understudied. This species of white-rot fungi was cultivated on solid-state media while under variable levels of known abiotic and biotic stressors to establish the effect upon fungal oil
Weronika Hewelt-Belka et al.
Food chemistry, 288, 154-161 (2019-03-25)
The complex nature of human breast milk (HBM) makes samples difficult to analyze, requiring several extraction techniques and analytical platforms to obtain high metabolome coverage. In this work, we combined liquid-liquid extraction (LLE) and solid-phase extraction (SPE) techniques to prepare
Claudia Glaser et al.
PloS one, 5(8), e12045-e12045 (2010-08-17)
Plasma fatty acid (FA) composition reflects dietary intake and endogenous turnover and is associated with health outcomes on a short and long term basis. The total plasma FA pool represents the composition of all FA containing lipid fractions. We developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service