Skip to Content
Merck
All Photos(1)

Key Documents

T7517

Sigma-Aldrich

Glyceryl tridecanoate

≥99% (GC)

Synonym(s):

1,2,3-Tricaprinoylglycerol, 1,2,3-Tridecanoylglycerol, Glycerol tricaprate, Glycerol tris(decanoate), Tricaprin, Tridecanoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)8COOCH2]2CHOCO(CH2)8CH3
CAS Number:
Molecular Weight:
554.84
Beilstein:
1717683
EC Number:
MDL number:
UNSPSC Code:
41141833
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99% (GC)

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC

InChI

1S/C33H62O6/c1-4-7-10-13-16-19-22-25-31(34)37-28-30(39-33(36)27-24-21-18-15-12-9-6-3)29-38-32(35)26-23-20-17-14-11-8-5-2/h30H,4-29H2,1-3H3

InChI key

LADGBHLMCUINGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erna Subroto et al.
Journal of food science and technology, 54(5), 1286-1292 (2017-04-19)
A series of experiments was conducted to determine optimum conditions for supercritical carbon dioxide extraction of candlenut oil. A Taguchi experimental design with L
Dewi van Harskamp et al.
Analytical and bioanalytical chemistry, 412(23), 5789-5797 (2020-07-11)
A new method for accurately analyzing octanoate enrichment in plasma was developed and validated. Samples were derivatized directly in plasma by transesterification with isobutanol and were analyzed by gas chromatography-mass spectrometry (GC-MS). This method was developed to analyze the precursor
Elisabetta Esposito et al.
Pharmaceutical research, 25(7), 1521-1530 (2008-01-04)
The present investigation describes a formulative study for the development of innovative drug delivery systems for bromocriptine. Solid lipid nanoparticles (SLN) based on different lipidic components have been produced and characterized. Morphology and dimensional distribution have been investigated by electron
Soo-Jeong Lim et al.
International journal of pharmaceutics, 243(1-2), 135-146 (2002-08-15)
Solid lipid nanoparticles (SLNs) have gained attention as a colloidal drug carrier, particularly for drugs with limited solubility. The poor aqueous solubility of all-trans retinoic acid (ATRA) has been a limiting factor in its clinical use. This study was undertaken
Doron J Aframian et al.
Quintessence international (Berlin, Germany : 1985), 41(3), e36-e42 (2010-03-10)
Salivary gland impairment is a major problem that can result in hyposalivation and a decrease in quality of life. Causes for mouth dryness can be grossly classified into three major groups: iatrogenic, immunogenic, and metabolic. At present, insufficient therapies exist

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service