Skip to Content
Merck
All Photos(1)

Key Documents

T2027

Sigma-Aldrich

Anti-Transferrin antibody produced in goat

whole antiserum

Synonym(s):

Anti-HEL-S-71p, Anti-PRO1557, Anti-PRO2086, Anti-TFQTL1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

indirect ELISA: 1:50,000
quantitative precipitin assay: 3.2-4.8 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TF(7018)

General description

Transferrin is a bilobed glycoprotein. The polypeptide chain of transferrin is composed of two homologous lobes i.e N- and C- lobes. Each lobe is made of two domains N1, N2 and C1, C2 which are linked by a hinge. It has two specific sites for binding of Fe3+.

Application

Anti-Transferrin antibody has been used:
  • in quantitative precipitin assay
  • in enzyme linked immunosorbent assay (ELISA)
  • in western blotting
  • in immunoblotting

Anti-Transferrin antibody produced in goat was used in western blotting analysis of HEK293 cell extracts.

Biochem/physiol Actions

Transferrin is the major iron transporter protein that binds Fe(+3). The transferring-iron import system consists of a TonB dependent transporter (TbpA), and a lipoprotein co-receptor (TbpB) and both proteins work together in the transport of iron across the membrane.
Transferrin plays a major role in providing iron as a source for haemoglobin synthesis. It binds to the transferrin receptor 1(TfR1).

Preparation Note

treated to remove lipoproteins

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Wang et al.
Journal of controlled release : official journal of the Controlled Release Society, 155(3), 386-392 (2011-07-16)
A proinsulin-transferrin (ProINS-Tf) recombinant fusion protein was designed and characterized for the sustained release of an active form of insulin (INS) by hepatoma cells. During incubation with H4IIE hepatoma cells, a gradual decline of ProINS-Tf concentration, with a concomitant generation
Receptor-mediated activation of a proinsulin-transferrin fusion protein in hepatoma cells.
Wang, Y., et al.
J. Controlled Release, 155, 386-392 (2011)
Nicholas Noinaj et al.
Molecular microbiology, 86(2), 246-257 (2012-09-11)
Two pathogenic species within the genus Neisseria cause the diseases gonorrhoea and meningitis. While vaccines are available to protect against four N. meningitidis serogroups, there is currently no commercial vaccine to protect against serogroup B or against N. gonorrhoeae. Moreover, the available
Hypoglycosylation is a common finding in antithrombin deficiency in the absence of a SERPINC 1 gene defect
de la Morena-Barrio M E, et al.
Journal of Thrombosis and Haemostasis, 14(8), 1549-1560 (2016)
New iron acquisition system in Bacteroidetes
Manfredi P, et al.
Infection and Immunity, 83(1), 300-310 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service