Skip to Content
Merck
All Photos(1)

Documents

SRP6273

Sigma-Aldrich

UPA human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

ATF, BDPLT5, PLAU, QPD, UPA, URK, Urokinase, u-PA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 45 kDa
observed mol wt 18 kDa
observed mol wt 32 kDa
observed mol wt 50 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... uPA(5328)

General description

DTT-reduced. Protein migrates as three bands corresponding to the long α chain, β chain and unprocessed full-length chain due to glycosylation and cleavage. Ser21, Ile179 & Lys156 is the predicted N-terminus.
The PLAU (plasminogen activator, urokinase) gene is mapped to human chromosome 10q22.2.
Urokinase-type plasminogen activator is also known as PLAU and uPA. The human uPA is initially synthesized as 431 amino acid precursor with an N-terminal signal peptide (20 residues). The single chain molecule is processed into a disulfide-linked two-chain molecule of different molecular weights. Two forms of the A chain exist, starting at Ser21 (the long form) and Lys156 (the short form). The long and short A chains are unique to the high and low molecular weight forms, respectively. The long A chain contains an EGF (epidermal growth factor)-like domain, responsible for binding of the PLAU receptor. The B chain corresponds to the catalytic domain. The gene encoding it is localized on human chromosome 10.

Biochem/physiol Actions

Urokinase-type plasminogen activator is a serine protease with extremely limited substrate specificity, cleaving the sequence Cys-Pro-Gly-Arg560-Val561-Val-Gly-Gly-Cys in plasminogen to form plasmin. It is a potent marker of invasion and metastasis in a variety of human cancers associated with breast, stomach, colon, bladder, ovary, brain and endometrium. uPA (plasminogen activator, urokinase) - mediated degradation of extracellular matrix of migrating cells is essential for tissue remodeling, invasiveness and angiogenesis. uPA promotes pathogenesis of Psoriasis and basal cell carcinoma. uPA activates growth factors and metalloproteinases.

Physical form

Lyophilized from 0.22 μm filtered solution in HEPES, NaCl and CaCl2. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increased expression of uPA, uPAR, and PAI-1 in psoriatic skin and in basal cell carcinomas.
Rubina K A, et al.
Archives of Dermatological Research, 309(6), 433-442 (2017)
Enhanced meta-analysis and replication studies identify five new psoriasis susceptibility loci.
Tsoi L C, et al.
Nature Communications, 6, 7001-7001 (2015)
Up-Regulation of PAI-1 and Down-Regulation of uPA Are Involved in Suppression of Invasiveness and Motility of Hepatocellular Carcinoma Cells by a Natural Compound Berberine.
Wang X
International Journal of Molecular Sciences, 17(4), 577-577 (2016)
Ajay Kumar et al.
Frontiers in immunology, 12, 807775-807775 (2022-01-04)
Leptospira, a zoonotic pathogen, is known to infect various hosts and can establish persistent infection. This remarkable ability of bacteria is attributed to its potential to modulate (activate or evade) the host immune response by exploiting its surface proteins. We
Jason J Paxman et al.
Nature communications, 10(1), 1967-1967 (2019-05-01)
Autotransporters are the largest family of outer membrane and secreted proteins in Gram-negative bacteria. Most autotransporters are localised to the bacterial surface where they promote colonisation of host epithelial surfaces. Here we present the crystal structure of UpaB, an autotransporter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service