Skip to Content
Merck
All Photos(1)

Documents

P7999

Sigma-Aldrich

Plasminogen from human plasma

≥2.0 units/mg protein, lumps and powder, suitable for streptokinase determination by clot formation procedure

Synonym(s):

Profibrinolysin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42030240
NACRES:
NA.54

biological source

human plasma

form

lumps and powder

specific activity

≥2.0 units/mg protein

mol wt

90 kDa

composition

protein, 2.0-7.0%

suitability

suitable for streptokinase determination by clot formation procedure

UniProt accession no.

foreign activity

plasmin ≤0.5% of base activity

storage temp.

2-8°C

Gene Information

human ... PLG(5340)

Looking for similar products? Visit Product Comparison Guide

General description

The plasminogen (PLG) gene is mapped to human chromosome 6q26. Plasminogen protein has a molecular weight of ?90kDa. It is composed of Pan-apple domain at the N-terminal, five kringle domains and serine protease domain at its C-terminal.

Application

Plasminogen is a single-chain glycoprotein found in human plasma and extracellular fluid. Certain activators, such as tissue plasminogen activator (tPA), convert plasminogen to its active form, plasmin. Plasminogen has been used to study its conversion into plasmin.
The enzyme from Sigma has been used to examine the ability of plasminogen in nasal secretion to promote S. aureus surface colonization. It has been used to assess the binding of recombinant OspC protein to plasminogen using enzyme-linked immunosorbent assay. It has also been used to screen FITC-labeled S. pneumoniae for protein interaction by solid-phase assay. In this assay; a well was coated with plasminogen and incubated with FITC-labeled bacteria.The fluorescence signal was read in a fluorimeter.

Biochem/physiol Actions

Plasminogen is the inactive precursor of the protease plasmin. Plasminogen is activated by the action of either tissue plasminogen activator (tPA), which primarily activates the fibrinolytic (thrombolytic) activity of plasmin, or urokinase plasminogen activator (uPA), which is associated with extracellular matrix remodeling and cell migration. Plasmin cleaves fibrin/fibrinogen and blood coagulation factors V/Va and VIII/VIIIa. It activates matrix metalloproteinases by cleaving the inactive proenzymes. It is also involved in the activation of some growth factors, such as vascular endothelial growth factor (VEGF) and transforming growth factor β (TGF-β).

Physical properties

Native-intact human plasminogen is a 791 amino acid glyco-protein with as many as 24 disulfide bonds. Plasminogen contains a single N-linked sialylated biantennary glycan. The two O-glycans possess a Gal β-1-3GalNAc core which are α-2-3 sialylated at the terminal Gal. An additional disialylated form has a second sialic acid residue with an α-2-6 linkage to GalNAc. Mono- and disialylated forms occur at a molar ratio of 80:20 in human plasminogen.

Unit Definition

One unit will produce one micromole of p-Nitroanilide from D-Val-Leu-Lys-p-Nitroanilide per minute at pH 7.5 at 37 °C. Activity is determined after activation to plasmin with urokinase.

Reconstitution

Solutions should be stored in buffer containing 20 mM lysine at neutral pH. The protein should be as concentrated as possible. Frozen in aliquots at −20 °C, it should be stable for several weeks.

Analysis Note

ε-Aminocaproic acid free
Plasma from each donor has been tested and found negative for antibody to HIV-1/HIV-2, antibody to HCV and HbSAg.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher G Earnhart et al.
Clinical and vaccine immunology : CVI, 18(6), 901-906 (2011-04-29)
Borrelia burgdorferi OspC is an outer membrane lipoprotein required for the establishment of infection in mammals. Due to its universal distribution among B. burgdorferi sensu lato strains and high antigenicity, it is being explored for the development of a next-generation
Genetic risk factors for portopulmonary hypertension in patients with advanced liver disease
Roberts KE, et al.
American Journal of Respiratory and Critical Care Medicine, 179(9), 835-842 (2009)
Jian Sha et al.
Journal of bacteriology, 191(9), 3095-3107 (2009-03-10)
In this study, we demonstrated that the surface-expressed enolase from diarrheal isolate SSU of Aeromonas hydrophila bound to human plasminogen and facilitated the latter's tissue-type plasminogen activator-mediated activation to plasmin. The bacterial surface-bound plasmin was more resistant to the action
Yan Wang et al.
Journal of cellular and molecular medicine, 24(16), 9255-9266 (2020-07-07)
Blood-brain barrier (BBB) integrity injury within the thrombolytic time window is becoming a critical target to reduce haemorrhage transformation (HT). We have previously reported that BBB damage was initially damaged in non-infarcted striatum after acute ischaemia stroke. However, the underlying
Cécile Frolet et al.
BMC microbiology, 10, 190-190 (2010-07-14)
Streptococcus pneumoniae is a widely distributed commensal Gram-positive bacteria of the upper respiratory tract. Pneumococcal colonization can progress to invasive disease, and thus become lethal, reason why antibiotics and vaccines are designed to limit the dramatic effects of the bacteria

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service