Skip to Content
Merck
All Photos(2)

Key Documents

SRP6156

Sigma-Aldrich

BMP-4 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

BMP-2B, Bone morphogenetic protein 2B, Bone morphogenetic protein 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

potency

2-10 ng/mL

mol wt

34 kDa (homodimer, glycosylated.)

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<1 EU/μg protein Endotoxin level (LAL test)

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BMP4(652)

General description

BMP4 (bone morphogenetic protein 4), a member of the highly conserved BMP family, is a growth factor and a pro-inflammatory and pro-atherogenic cytokine. BMPs are part of the TGFβ (tumor growth factor) superfamily of proteins. BMP4 gene is localized to human chromosome 14q22-q23.

Biochem/physiol Actions

BMP4 (bone morphogenetic protein 4) is involved in modulating cell development in a wide variety of cells. It promotes the proliferation and migration of hepatocellular carcinoma cells, and the multiplication of ALK3 (activin-like kinase)-positive cells in skeletal muscle. This protein prevents the proliferation of lung fibroblasts and induces their differentiation into myocytes. In Sertoli cells, it promotes DNA synthesis and hence, the proliferation of Sertoli cells through Smad1/5 (small mothers against decapentaplegic) and ID2/3 (DNA binding protein inhibitor) pathway, and aberration in BMP4 expression results in azoospermia. In human males, the serum levels of BMP4 is linked with coronary artery disease (CAD) severity. BMP4 is crucial for maintaining the self-renewal capacity of ES (embryonic stem) cells, and achieves so by inhibiting extracellular receptor kinase (ERK) and p38 mitogen-activated protein kinase (MAPK) pathways.

Physical form

Lyophilized from a PBS solution.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Reconstitute in sterile 4 mM HCl containing 0.1% endotoxin-free recombinant human serum albumin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BMP4 promotes human Sertoli cell proliferation via Smad1/5 and ID2/3 pathway and its abnormality is associated with azoospermia.
Hai Y et al
Discovery Medicine, 19(105), 311-325 (2015)
Bmp4 is required for the generation of primordial germ cells in the mouse embryo.
Lawson KA et al
Genes & Development, 13(4), 424-436 (1999)
BMP4 supports self-renewal of embryonic stem cells by inhibiting mitogen-activated protein kinase pathways.
Qi X et al
Proceedings of the National Academy of Sciences of the USA, 101(16), 6027-6032 (2004)
Bmp4 and Fgf10 play opposing roles during lung bud morphogenesis.
Weaver M et al
Development, 127(12), 2695-2704 (2000)
Serum Bone Morphogenic Protein-4 Contributes to Discriminating Coronary Artery Disease Severity.
Park CS et al
Medicine, 94(3), e1530-e1530 (2015)

Articles

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service