Skip to Content
Merck
All Photos(8)

Key Documents

H3149

Sigma-Aldrich

Heparin sodium salt from porcine intestinal mucosa

Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

biological source

Porcine intestinal mucosa

Quality Level

type

Grade I-A

product line

BioReagent

form

powder

specific activity

≥180 USP units/mg

technique(s)

analytical sample preparation: suitable
cell culture | mammalian: suitable

color

beige

solubility

H2O: 50 mg/mL

application(s)

cell analysis
diagnostic assay manufacturing
life science and biopharma
sample preparation

SMILES string

[Na+].[S](=O)(=O)(NC1C(OC(C(C1O[S](=O)(=O)O)OC4OC(C(C(C4O[S](=O)(=O)O)O)O)C(=O)O)CO)OC2C(OC(C(C2O)O[S](=O)(=O)O)OC3C(OC(C(C3O)NC(=O)C)O)CO[S](=O)(=O)O)C(=O)O)O

InChI

1S/C26H42N2O37S5.Na/c1-4(30)27-7-9(31)13(6(56-23(7)39)3-55-67(43,44)45)58-26-19(65-70(52,53)54)12(34)16(20(62-26)22(37)38)60-24-8(28-66(40,41)42)15(63-68(46,47)48)14(5(2-29)57-24)59-25-18(64-69(49,50)51)11(33)10(32)17(61-25)21(35)36;/h5-20,23-26,28-29,31-

InChI key

JRTRSJGZMRQDHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heparin is mainly responsible for the delay in the coagulation of blood. It enhances the antithrombin-mediated inactivation of proteases in the coagulation pathway.
Heparin sodium, a sulfated polysaccharide from the glycosaminoglycan family, plays a vital role in cell culture, cancer biology, and drug discovery research. Its impact on protein and cell interactions makes it valuable for studying molecular and cellular processes, tumor modeling, exploring binding interactions, and as a cell culture supplement to support cell viability.

Application

  • Heparin sodium salt from porcine intestinal mucosa has been used in sLRP (soluble circulating low density lipoprotein receptor-related protein-1) purification from human plasma.
  • It has been used as a supplement in M199 medium for culturing of human umbilical vein endothelial cells.
  • It has been used as an anticoagulant agent during isolation of blood.

Features and Benefits

  • High-purity compound suitable for a wide variety of research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Factor Xa and thrombin, but not factor VIIa, elicit specific cellular responses in dermal fibroblasts.
Bachli EB et al.
Journal of Thrombosis and Haemostasis, 1, 1935-1935 (2003)
Structure of the antithrombin-binding site in heparin.
Lindahl U et al.
Proceedings of the National Academy of Sciences of the USA, 76, 3198-3202 (1979)
Effects of T-lymphocyte depletion on muscle fibrosis in the mdx mouse.
Morrison J, et al.
The American Journal of Pathology, 166, 1701-1701 (2005)
Mohamed Abdouh et al.
Journal of experimental & clinical cancer research : CR, 36(1), 113-113 (2017-09-01)
Horizontal transfer of malignant traits from the primary tumor to distant organs, through blood circulating factors, has recently become a thoroughly studied metastatic pathway to explain cancer dissemination. Recently, we reported that oncosuppressor gene-mutated human cells undergo malignant transformation when
Keisuke Yoshida et al.
Nature communications, 9(1), 3885-3885 (2018-09-27)
The majority of histones are replaced by protamines during spermatogenesis, but small amounts are retained in mammalian spermatozoa. Since nucleosomes in spermatozoa influence epigenetic inheritance, it is important to know how histones are distributed in the sperm genome. Conflicting data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service