Skip to Content
Merck
All Photos(1)

Key Documents

SRP0406

Sigma-Aldrich

Histone H2a full length human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

H2AFQ, histone cluster 2, H2ac

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

14.8 kDa

packaging

pkg of 1 mg

storage condition

avoid repeated freeze/thaw cycles

concentration

1.2 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

Histone cluster 2 H2A family member c (HIST2H2AC) is located on human chromosome 1q21. It is usually present in undifferentiated/proliferating cells. This canonical histone isoform has a protruding tail at both the N- and C-termini.
Histones make up the nucleosome, which is the fundamental unit of the chromatin. Each nucleosome consists of 146 base pairs of DNA which is wrapped around an octamer of four core histones H2A, H2B, H3 and H4. Histone cluster 2 H2A family member c (HIST2H2AC) has many variants. Human Histone 2A (GenBank Accession No. NM_033445) amino acids 2-130 (end) with N-terminal His-tag, MW = 14.8 kDa, expressed in an E. coli expression system.

Biochem/physiol Actions

Histone cluster 2 H2A family member c (HIST2H2AC) has a role in maintaining the structure of the chromatin.
Histone cluster 2 H2A family member c (HIST2H2AC) participates in multiple epigenetic regulations and carcinogenesis. It also participates in embryonic stem (ES) cell biology and regulates the expression of genes. It modulates proliferation and epithelial-mesenchymal transition in mammary epithelial and breast cancer cells. Hist2h2ac also controls epithelial differentiation.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The histone H2A isoform Hist2h2ac is a novel regulator of proliferation and epithelial-mesenchymal transition in mammary epithelial and in breast cancer cells
Monteiro FL, et al.
Cancer Letters, 396, 42-52 (2017)
Wenlai Zhou et al.
The international journal of biochemistry & cell biology, 41(1), 12-15 (2008-10-22)
The precise molecular strategies that coordinate patterns of transcriptional response to specific signals is central for understanding normal development and disease. Precise control of transcriptional programs underlying metazoan development is modulated by enzymatically active coregulatory complexes, coupled with epigenetic strategies.
Truncation of histone H2A's C-terminal tail, as is typical for Ni(II)-assisted specific peptide bond hydrolysis, has gene expression altering effects.
Karaczyn AA
Annals of Clinical and Laboratory Science, 39(3), 251-262 (2009)
Acetylation of vertebrate H2A.Z and its effect on the structure of the nucleosome.
Ishibashi T
Biochemistry, 48(22), 5007-5017 (2009)
Mary Ann Osley
Briefings in functional genomics & proteomics, 5(3), 179-189 (2006-06-15)
Histone ubiquitylation has emerged as an important chromatin modification with roles in transcription and trans-histone methylation. In the past several years, there has been dramatic progress in the identification of factors that control ubiquitin attachment to the core histones H2A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service