Skip to Content
Merck
All Photos(3)

Documents

H5505

Sigma-Aldrich

Histone from calf thymus

Type III-S

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1895465
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

biological source

bovine (calf) thymus

type

Type III-S

form

powder

concentration

≥60% protein (biuret)

technique(s)

activity assay: suitable

solubility

H2O: soluble 10 mg/mL, clear to hazy, colorless to light yellow

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Histone is suitable for use:
  • as a positive control in the in vitro kinase assay to examine the ability of Cdc2-cyclin B1 to phosphorylate GST-CD-4 protein
  • in Cdk5 activity assay
  • in histone H1 kinase assay
  • as a Poly(ADP-ribose) (PAR) acceptor in the assessment of in vitro PAR polymerase activity
  • as an acceptor of 32P from [γ-32P]ATP in order to measure protein kinase activity of lysate from small and large luteal cells

Biochem/physiol Actions

Histones are a group of proteins characterized by high levels of lysine and arginine that form reversible complexes with DNA. Based on the fractions, the molar mass of histones varies between 11-21kDa. Five different fractions have been isolated and characterized. They include H1 (lysine rich), H2a and H2b that are slightly lysine rich, H3 and H4 that are arginine rich. H1 acts as a link between "beads" (nu bodies) on the chromatin chain.

Features and Benefits

Do not confuse Sigma "Type" designations with the "Fraction" designations used by Luck and co-workers.

Other Notes

Characterized as mainly subgroup f1 by SDS-PAGE

Preparation Note

Lysine-rich fraction as isolated and described by de Nooij, E. and Westenbrink, H.G., Biochim. Biophys. Acta, 62, 608 (1962).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Scott T
Concise encyclopedia of biochemistry, 265-265 (1988)
Anne Cecília Nascimento da Cruz et al.
Pharmaceuticals (Basel, Switzerland), 12(4) (2019-11-21)
Fourteen arylsemicarbazone derivatives were synthesized and evaluated in order to find agents with potential anticancer activity. Cytotoxic screening was performed against K562, HL-60, MOLT-4, HEp-2, NCI-H292, HT-29 and MCF-7 tumor cell lines. Compounds 3c and 4a were active against the
Q P Dou et al.
Cancer research, 53(7), 1493-1497 (1993-04-01)
Progression of cells into S phase is proposed to be determined by accumulation of a labile protein (the restriction point protein R; A. B. Pardee, Proc. Natl. Acad. Sci. USA, 71: 1286-1290, 1974). We report here that cyclin E and
P Balsas et al.
Journal of hematology & oncology, 10(1), 80-80 (2017-04-01)
Pharmacological inhibition of B cell receptor (BCR) signaling has recently emerged as an effective approach in a wide range of B lymphoid neoplasms. However, despite promising clinical activity of the first Bruton's kinase (Btk) and spleen tyrosine kinase (Syk) inhibitors
Mária Čížková et al.
Cells, 8(7) (2019-07-20)
DNA damage is a ubiquitous threat endangering DNA integrity in all living organisms. Responses to DNA damage include, among others, induction of DNA repair and blocking of cell cycle progression in order to prevent transmission of damaged DNA to daughter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service