Skip to Content
Merck
All Photos(2)

Documents

SML3799

Sigma-Aldrich

Nilotinib

≥98% (HPLC)

Synonym(s):

4-Methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-N-[5-(4-methyl-1H-imidazol-1-yl)-3-(trifluoromethyl)phenyl]benzamide, 4-Methyl-N-[3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-benzamide, AMN 107, AMN107

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22F3N7O
CAS Number:
Molecular Weight:
529.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Nilotinib (AMN107) is an orally available, selective and potent ATP-competitive wild-type and mutant Bcr-Abl kinase inhibitor that is 10-30-fold more potent than imatinib. Nilotinib is used to treat Philadelphia chromosome (Ph+)-positive chronic myelogenous leukemia (CML). Nilotinib reduces midbrain Bcr-Abl autophosphorylation, amyloid-β levels, and neuronal loss, as well as improves autophagosome clearance and reverses cognitive deficits in the Tg2576 transgenic mouse model of Alzheimer’s disease.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - STOT RE 1

Target Organs

Liver,Kidney,gallbladder

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of AMN107, a selective inhibitor of native and mutant Bcr-Abl
Cancer Cell, 7(2), 129-141 (2005)
Tomer Meirson et al.
Oncotarget, 9(31), 22158-22183 (2018-05-19)
Metastatic dissemination of cancer cells from the primary tumor and their spread to distant sites in the body is the leading cause of mortality in breast cancer patients. While researchers have identified treatments that shrink or slow metastatic tumors, no
Livia La Barbera et al.
Progress in neurobiology, 202, 102031-102031 (2021-03-09)
What happens precociously to the brain destined to develop Alzheimer's Disease (AD) still remains to be elucidated and this is one reason why effective AD treatments are missing. Recent experimental and clinical studies indicate that the degeneration of the dopaminergic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service