Skip to Content
Merck
All Photos(1)

Key Documents

SML3228

Sigma-Aldrich

Rimiducid

≥90% (HPLC)

Synonym(s):

(S,2S,2′S)-((1R,1′R)-1,1′-(3,3′-(2,2′-(Ethane-1,2-diylbis(azanediyl))bis(2-oxoethane-2,1-diyl))bis(oxy)bis(3,1-phenylene))bis(3-(3,4-dimethoxyphenyl)propane-1,1-diyl)) bis(1-((S)-2-(3,4,5-trimethoxyphenyl)butanoyl)piperidine-2-carboxylate), 1,1′-{Ethylenebis[azanediyl(2-oxoethane-2,1-diyl)oxy-3,1-phenylene]}bis[(1R)-3(3,4-dimethoxyphenyl)propyl]bis{(2S)-1-[(2S)-2-(3,4,5trimethoxyphenyl)butanoyl]piperidine-2-carboxylate}, 2,2′-[1,2-Ethanediylbis[imino(2-oxo-2,1-ethanediyl)oxy-3,1-phenylene[(1R)-3-(3,4-dimethoxyphenyl)propylidene]]] bis[(2S)-1-[(2S)-1-oxo-2-(3,4,5-trimethoxyphenyl)butyl]-2-piperidinecarboxylate], AP 1903, AP-1903, AP1903

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H98N4O20
CAS Number:
Molecular Weight:
1411.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

N2([C@@H](CCCC2)C(=O)O[C@H](CCc8cc(c(cc8)OC)OC)c3cc(ccc3)OCC(=O)NCCNC(=O)COc4cc(ccc4)[C@H](OC(=O)[C@H]6N(CCCC6)C(=O)[C@@H](CC)c7cc(c(c(c7)OC)OC)OC)CCc5cc(c(cc5)OC)OC)C(=O)[C@@H](CC)c1cc(c(c(c1)OC)OC)OC

InChI key

GQLCLPLEEOUJQC-ZTQDTCGGSA-N

Biochem/physiol Actions

High-affinity FKBP12 F36V mutant cross-linker for Chemical Induction of Dimerization (CID) of Chimeric Antigen Receptor (CAR) in vitro and in vivo.
Rimiducid (AP1903) is a homodimeric FKBP12 Phe36Val (F36V-FKBP, FKBP-F36V) mutant-selective cross-linker composed of two high-affinity ligands, each with 713-fold selectivity over endogenous FKBP (Kd = 94 pM/F36V- vs 67 nM/Wt-FKBP using 4′-AF-labeled rimiducid). Rimiducid activates signaling events via Chemical Induction of Dimerization (CID) by cross-linking FKBP12-F36 fusions (Chimeric Antigen Receptor or CAR) expressed in cells in cultures (apoptosis induction EC50 = 0.1 nM; HT1080 expressing FKBP(F36V)-Fas intracellular domain) and in animals in vivo (EC50 = 0.4 mg/kg i.v. in a murine model of conditional cell ablation).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Berger et al.
Blood, 103(4), 1261-1269 (2003-10-18)
Conditional suicide genes derived from pathogens have been developed to confer drug sensitivity and enhance safety of cell therapy, but this approach is limited by immune responses to the transgene product. We examined a strategy to regulate survival of transferred
Xiaoou Zhou et al.
Blood, 125(26), 4103-4113 (2015-05-16)
To test the feasibility of a single T-cell manipulation to eliminate alloreactivity while sparing antiviral and antitumor T cells, we infused 12 haploidentical hematopoietic stem cell transplant patients with increasing numbers of alloreplete haploidentical T cells expressing the inducible caspase
Dongpeng Jiang et al.
Leukemia, 34(3), 821-830 (2019-10-19)
CD19-redirected CAR-T immunotherapy has emerged as a promising strategy for treatment of B cell lymphoma, however, many patients often relapsed due to antigen loss. Therefore, it is urgently needed to explore other suitable antigens targeted by CAR-T cells to cure
D C Thomis et al.
Blood, 97(5), 1249-1257 (2001-02-27)
Graft-versus-host disease (GVHD) is a major complication of allogeneic bone marrow transplantation. One strategy to treat GVHD is to equip donor T cells with a conditional suicide mechanism that can be triggered when GVHD occurs. The herpes simplex virus thymidine
Xiaomei Wang et al.
Blood advances, 4(9), 1950-1964 (2020-05-10)
Natural killer (NK) cells expressing chimeric antigen receptors (CARs) are a promising anticancer immunotherapy, leveraging both innate NK cell antitumor activity and target-specific cytotoxicity. Inducible MyD88/CD40 (iMC) is a potent, rimiducid-regulated protein switch that has been deployed previously as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service