Skip to Content
Merck
All Photos(1)

Key Documents

SML3137

Sigma-Aldrich

Boceprevir

≥98% (HPLC)

Synonym(s):

(1R,2S,5S)-N-[3-Amino-1-(cyclobutylmethyl)-2,3-dioxopropyl]-3-[(2S)-2-[[[(1,1-dimethylethyl)amino]carbonyl]amino]-3,3-dimethyl-1-oxobutyl]-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxamide, (1R,5S)-N-[3-Amino-1-(cyclobutylmethyl)-2,3-dioxopropyl]-3-[2(S)-[[[(1,1-dimethylethyl)amino]carbonyl]amino]-3,3-dimethyl-1-oxobutyl]-6,6-dimethyl-3-azabicyclo[3.1.0]hexan-2(S)-carboxamide, SCH 503034, SCH503034

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H45N5O5
CAS Number:
Molecular Weight:
519.68
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1C[C@@]([H])([C@]2([C@]1(C(NC(CC3CCC3)C(C(N)=O)=O)=O)[H])[H])C2(C)C)[C@](C(C)(C)C)(NC(NC(C)(C)C)=O)[H]

InChI

1S/C27H45N5O5/c1-25(2,3)20(30-24(37)31-26(4,5)6)23(36)32-13-15-17(27(15,7)8)18(32)22(35)29-16(19(33)21(28)34)12-14-10-9-11-14/h14-18,20H,9-13H2,1-8H3,(H2,28,34)(H,29,35)(H2,30,31,37)/t15-,16?,17-,18-,20+/m0/s1

InChI key

LHHCSNFAOIFYRV-DOVBMPENSA-N

Biochem/physiol Actions

Boceprevir is an orally bioavailable, potent and selective hepatitis C virus (HCV) NS3 protease inhibitor (Ki = 14 nM; cell-based replicon assay IC50 = 350 nM).

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunlong Ma et al.
Cell research, 30(8), 678-692 (2020-06-17)
A new coronavirus SARS-CoV-2, also called novel coronavirus 2019 (2019-nCoV), started to circulate among humans around December 2019, and it is now widespread as a global pandemic. The disease caused by SARS-CoV-2 virus is called COVID-19, which is highly contagious
B A Malcolm et al.
Antimicrobial agents and chemotherapy, 50(3), 1013-1020 (2006-02-24)
Cleavage of the hepatitis C virus (HCV) polyprotein by the viral NS3 protease releases functional viral proteins essential for viral replication. Recent studies by Foy and coworkers strongly suggest that NS3-mediated cleavage of host factors may abrogate cellular response to
Yanmei Hu et al.
bioRxiv : the preprint server for biology (2020-11-04)
As the COVID-19 pandemic continues to fold out, the morbidity and mortality are increasing daily. Effective treatment for SARS-CoV-2 is urgently needed. We recently discovered four SARS-CoV-2 main protease (Mpro) inhibitors including boceprevir, calpain inhibitors II and XII and GC-376
Andrew J Prongay et al.
Journal of medicinal chemistry, 50(10), 2310-2318 (2007-04-21)
The structures of both the native holo-HCV NS3/4A protease domain and the protease domain with a serine 139 to alanine (S139A) mutation were solved to high resolution. Subsequently, structures were determined for a series of ketoamide inhibitors in complex with
Xiao Tong et al.
Antiviral research, 77(3), 177-185 (2008-01-19)
An issue of clinical importance in the development of new antivirals for HCV is emergence of resistance. Several resistance loci to ketoamide inhibitors of the NS3/4A protease have been identified (residues V36, T54, R155, A156, and V170) by replicon and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service