Skip to Content
Merck
All Photos(1)

Key Documents

Y0000744

Dextranomer

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dextranomer

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dextranomer EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Injection therapy for reflux: why it works and why it fails.
Andrew J Kirsch
The Journal of urology, 188(1), 16-17 (2012-05-16)
R Angioli et al.
Minerva ginecologica, 60(6), 543-550 (2008-11-05)
Urinary incontinence consist in voluntary urine leakage. Female affected in the world are about 200 thousand. Urinary incontinence affect severely women quality of life. There are different kinds of urinary incontinence that can be treated in different ways. We can
Oliver Schwandner et al.
Surgical innovation, 18(2), 130-135 (2011-01-20)
The aim of this prospective study was to analyze safety and functional outcome of transanal submucosal injection of dextranomer hyaluronic acid ("bulking agents therapy") in patients with passive fecal incontinence. All patients who underwent transanal injection therapy were prospectively enrolled
Editorial comment.
Anthony A Caldamone
The Journal of urology, 188(1), 261-261 (2012-05-19)
Jonathan F Kalisvaart et al.
Journal of pediatric urology, 8(4), 359-365 (2011-08-09)
Follow-up of patients undergoing dextranomer/hyaluronic acid injection for vesico-ureteral reflux (VUR) is controversial. The purpose of our study was to test the hypothesis that patients undergoing the double hydrodistention-implantation technique (Double HIT) have a higher clinical and radiographic success rate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service