Skip to Content
Merck
All Photos(1)

Key Documents

SML3049

Sigma-Aldrich

AMG PERK 44

≥98% (HPLC)

Synonym(s):

4-(2-Amino-4-methyl-3-(2-methylquinolin-6-yl)benzoyl)-1-methyl-2,5-diphenyl-1H-pyrazol-3(2H)-one hydrochloride, 4-[2-Amino-4-methyl-3-(2-methyl-6-quinolinyl)benzoyl]-1,2-dihydro-1-methyl-2,5-diphenyl-3H-pyrazol-3-one hydrochloride, AMG′44, AMG44, Compound 44, PERK inhibitor 44, PERK-IN-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H28N4O2 · xHCl
CAS Number:
Molecular Weight:
524.61 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1N(N(C(C2=CC=CC=C2)=C1C(C3=CC=C(C(C4=CC=C5N=C(C=CC5=C4)C)=C3N)C)=O)C)C6=CC=CC=C6.[xHCl]

Biochem/physiol Actions

AMG PERK 44 (AMG44) is an orally active (F = 46% (mouse) & 55% (rat) post 2.5 mg/kg po.), active site-targeing, potent and highly selective PERK (EIF2AK3) inhibitor (IC50 = 6 nM; GCN2 IC50 = 7.3 μM, b-Raf IC50 >1 μM, <65% inhibition at 1 μM against 387 kinases). AMG44 inhibits cellular PERK autophosphorylation (pT980 IC50 = 84 nM; HT1080 T-REx-PERK-FLAG transfectants) and ER stress-induced bulk protein synthesis (IC50 = 400 nM; U2OS), as well as PERK pT980 in HT1080 Matrigel plug implant in mice in vivo (ED50 = 3 mg/kg & ED90 = 60 mg/kg 4 hr post po.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia M Peñaranda-Fajardo et al.
Cell death & disease, 10(10), 690-690 (2019-09-20)
Patients with aggressive brain tumors, named glioblastoma multiforme (GBM), have a poor prognoses. Here we explored if the ER stress/unfolded protein response (UPR) is involved in the pathophysiology of GBM and may provide novel therapeutic targets. Immunohistochemical analyses of a
Adrian L Smith et al.
Journal of medicinal chemistry, 58(3), 1426-1441 (2015-01-15)
The structure-based design and optimization of a novel series of selective PERK inhibitors are described resulting in the identification of 44 as a potent, highly selective, and orally active tool compound suitable for PERK pathway biology exploration both in vitro
Manar G Helal et al.
Life sciences, 239, 117017-117017 (2019-11-05)
Saxagliptin (Saxa), a dipeptidyl dipeptidase-4 (DPP-4) inhibitor, is widely used for the treatment of type 2 diabetes mellitus. It has been documented to have immunomodulatory and anti-inflammatory actions. Our objective was to delineate the protective effect and the underlying mechanism
Diego Rojas-Rivera et al.
Cell death and differentiation, 24(6), 1100-1110 (2017-04-30)
Accumulation of unfolded proteins in the endoplasmic reticulum (ER) causes a state of cellular stress known as ER stress. The cells respond to ER stress by activating the unfolded protein response (UPR), a signaling network emerging from the ER-anchored receptors
Mohamed Mahameed et al.
Cell death & disease, 10(4), 300-300 (2019-04-02)
IRE1, PERK, and ATF6 are the three transducers of the mammalian canonical unfolded protein response (UPR). GSK2606414 is a potent inhibitor of PERK, while KIRA6 inhibits the kinase activity of IRE1. Both molecules are frequently used to probe the biological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service