Skip to Content
Merck
All Photos(3)

Key Documents

SML0843

Sigma-Aldrich

ISRIB

≥98% (HPLC), powder, integrated stress response inhibitor

Synonym(s):

Integrated Stress Response inhibitor, trans-N,N′-(Cyclohexane-1,4-diyl)bis(2-(4-chlorophenoxy)acetamide, trans-N,N′-1,4-Cyclohexanediylbis[2-(4-chlorophenoxy)-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24Cl2N2O4
CAS Number:
Molecular Weight:
451.34
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

ISRIB, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC1=CC=C(OCC(N[C@@H]2CC[C@@H](NC(COC3=CC=C(Cl)C=C3)=O)CC2)=O)C=C1

InChI

1S/C22H24Cl2N2O4/c23-15-1-9-19(10-2-15)29-13-21(27)25-17-5-7-18(8-6-17)26-22(28)14-30-20-11-3-16(24)4-12-20/h1-4,9-12,17-18H,5-8,13-14H2,(H,25,27)(H,26,28)/t17-,18-

InChI key

HJGMCDHQPXTGAV-IYARVYRRSA-N

Application

ISRIB (integrated stress response inhibitor) has been used as an inhibitor of integrated stress response.[1][2]

Biochem/physiol Actions

ISRIB (integrated stress response inhibitor) as a drug, imparts resistance to the cells against the downstream effects of eukaryotic initiation factor 2 (eIF2)α phosphorylation such as activating transcription factor 4 (ATF4) and eukaryotic translation initiation factor 4E (eIF4E)-binding protein 1 (4E-BP1) induction.[1]
ISRIB is a blood- brain barrier penetrant, potent and selective inhibitor of integrated stress response (ISR) that impairs adaptation to ER stress. ISRIB is a potent inhibitor of PERK signaling, which potently reverses the effects of eIF2a phosphorylation and restores the cells translation capacity. ISRIB enhances cognitive memory in rodents.
ISRIB is a blood- brain barrier penetrant, potent and selective inhibitor of the inhibitor of PERK signaling; cognitive memory enhancer..

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Balakrishnan et al.
Biochimica et biophysica acta. Molecular basis of disease, 1865(11), 165516-165516 (2019-07-31)
Loss of galactose-1 phosphate uridylyltransferase (GALT) activity in humans results in Classic Galactosemia, and the GalT-deficient (GalT-/-) mouse mimics the patient condition. GalT-/- ovaries display elevated endoplasmic reticulum (ER) stress marker, BiP, and downregulated canonical phosphatidylinositol 3-kinase (Pi3k)/protein kinase B
Z D Kabir et al.
Molecular psychiatry, 22(8), 1096-1109 (2017-06-07)
CACNA1C, encoding the Ca
Caitlin M Rodriguez et al.
Nature neuroscience, 23(3), 386-397 (2020-02-19)
Repeat-associated non-AUG-initiated translation of expanded CGG repeats (CGG RAN) from the FMR1 5'-leader produces toxic proteins that contribute to neurodegeneration in fragile X-associated tremor/ataxia syndrome. Here we describe how unexpanded CGG repeats and their translation play conserved roles in regulating
Philippe Fernandes et al.
Communications biology, 4(1), 127-127 (2021-01-31)
Common fragile sites (CFSs) are genomic regions frequently involved in cancer-associated rearrangements. Most CFSs lie within large genes, and their instability involves transcription- and replication-dependent mechanisms. Here, we uncover a role for the mitochondrial stress response pathway in the regulation
Ye-Jin Park et al.
International journal of molecular sciences, 21(15) (2020-08-01)
Stress granules are membraneless organelles composed of numerous components including ribonucleoproteins. The stress granules are characterized by a dynamic complex assembly in response to various environmental stressors, which has been implicated in the coordinated regulation of diverse biological pathways, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service