Skip to Content
Merck
All Photos(1)

Key Documents

SML1278

Sigma-Aldrich

Geldanamycin Ready Made Solution

1 mg/mL, from Streptomyces hygroscopicus

Synonym(s):

2-Azabicyclo[16.3.1]docosa-4,6,10,18,21-pentaene-3,20,22-trione, 9,13-dihydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-, 9-carbamate (8CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H40N2O9
CAS Number:
Molecular Weight:
560.64
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

Streptomyces hygroscopicus

Quality Level

form

DMSO solution

storage condition

protect from light

concentration

1 mg/mL

antibiotic activity spectrum

neoplastics

Mode of action

enzyme | inhibits

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C29H40N2O9/c1-15-11-19-25(34)20(14-21(32)27(19)39-7)31-28(35)16(2)9-8-10-22(37-5)26(40-29(30)36)18(4)13-17(3)24(33)23(12-15)38-6/h8-10,13-15,17,22-24,26,33H,11-12H2,1-7H3,(H2,30,36)(H,31,35)/b10-8-,16-9+,18-13+/t15-,17+,22+,23+,24-,26+/m1/s1

InChI key

QTQAWLPCGQOSGP-KSRBKZBZSA-N

General description

Chemical structure: benzenoid

Application

Geldanamycin Ready-Made Solution has been used as a heat shock protein 90 (Hsp90) inhibitor to pre-treat proteosome holo-enzyme during proteasome assay to study the role of proteasome on fibril fragmentation.

Biochem/physiol Actions

Geldanamycin is a benzoquinone ansamycin and is a well known tyrosine kinase inhibitor. It possesses anticancer effects and shows therapeutic effects against osteosarcoma.
Geldanamycin is a potent antitumor antibiotic active at nanomolar concentration against 60 cell lines. Geldanamycin binds specifically to the heat shock protein Hsp90 and to its endoplasmic reticulum homologue GP96, and thus interferes with conformational maturation of proteins and the cellular stress response. In addition, Geldanamycin is a potent inhibitor of the nuclear hormone receptor family.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Magdalena Gorska et al.
PloS one, 8(8), e71135-e71135 (2013-09-10)
Osteosarcoma is one of the most malignant tumors of childhood and adolescence that is often resistant to standard chemo- and radio-therapy. Geldanamycin and geldanamycin analogs have been recently studied as potential anticancer agents for osteosarcoma treatment. Here, for the first
Rachel Cliffe et al.
Cell reports, 26(8), 2140-2149 (2019-02-21)
Filamentous aggregates (fibrils) are regarded as the final stage in the assembly of amyloidogenic proteins and are formed in many neurodegenerative diseases. Accumulation of aggregates occurs as a result of an imbalance between their formation and removal. Here we use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service