Skip to Content
Merck
All Photos(4)

Key Documents

R2146

Sigma-Aldrich

Radicicol from Diheterospora chlamydosporia

greener alternative

solid

Synonym(s):

Monorden, [1aS-(1aR*,2Z,4E,14*,15aR*)]-8-Chloro-1a,14,15,15a-tetrahydro-9,11-dihydroxy-14-methyl-6H-oxireno[e][2]benzoxacyclotetradecin-6,12(7H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17ClO6
CAS Number:
Molecular Weight:
364.78
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

greener alternative product score

old score: 15
new score: 9
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Atom Economy
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white to yellow-white

solubility

ethanol: 10 mg/mL

antibiotic activity spectrum

fungi

greener alternative category

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

C[C@@H]1C[C@H]2O[C@@H]2C=C\C=C\C(=O)Cc3c(Cl)c(O)cc(O)c3C(=O)O1

InChI

1S/C18H17ClO6/c1-9-6-15-14(25-15)5-3-2-4-10(20)7-11-16(18(23)24-9)12(21)8-13(22)17(11)19/h2-5,8-9,14-15,21-22H,6-7H2,1H3/b4-2+,5-3-/t9-,14-,15-/m1/s1

InChI key

WYZWZEOGROVVHK-GTMNPGAYSA-N

Gene Information

Application

Radicicol from Diheterospora chlamydosporia has been used as an inhibitor of heat shock protein 90 (Hsp90):
  • to study its effects on lifespan extension and health in Caenorhabditis elegans
  • to study its effects on protein aggregation in yeast
  • to study its effects on xanthone sensitized cancer cells

Biochem/physiol Actions

Radicicol is an antifungal macrolactone antibiotic that is found in Diheterospora chlamydosporia, Chaetomium chiversii, and Monosporium bonorden. It functions as an inhibitor of tyrosine kinase and heat shock protein 90 (Hsp90). Radicicol is involved in the suppression of transformation of various proto-oncogenes such as Ras, Mos, and Src. It also suppresses the activity of mitogen-induced cyclooxygenase-2 (COX-2) and phosphoinositide-dependent kinase 1 (PDK1). Radicicol is involved in arresting the cell cycle at the G1-S phase. It exhibits ant-cancer and anti-angiogenic activity in vivo.

Caution

Photosensitive

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Muta. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S V Sharma et al.
Oncogene, 16(20), 2639-2645 (1998-06-19)
Radicicol, a macrocyclic anti-fungal antibiotic, has the ability to suppress transformation by diverse oncogenes such as Src, Ras and Mos. Despite this useful property, the mechanism by which radicicol exerts its anti-transformation effects is currently unknown. To understand the transformation-suppressing
Peter W Piper et al.
Open biology, 2(12), 120138-120138 (2012-12-29)
Heat shock protein 90 (Hsp90) is a promising cancer drug target as a molecular chaperone critical for stabilization and activation of several of the oncoproteins that drive cancer progression. Its actions depend upon its essential ATPase, an activity fortuitously inhibited
Targeting DNA topoisomerases in parasitic protozoa by natural products: Chemical and biological perspectives
Chowdhury S, et al.
Studies in Natural Products Chemistry, 67, 389-410 (2021)
Yilin Zhao et al.
The Journal of surgical research, 182(2), 312-318 (2012-11-10)
Intestinal injury is a key feature in sepsis. Inhibitors of heat shock protein 90 (Hsp90) have been shown to exert protective effects in models of inflammation. Herein, we hypothesized that Hsp90 might regulate intestinal inflammation and leakage in abdominal sepsis.
Yonghan He et al.
Biochemical and biophysical research communications, 436(2), 169-174 (2013-06-04)
Heat shock protein 90 (Hsp90) is involved in various cellular processes, such as cell proliferation, differentiation and apoptosis. As adipocyte differentiation plays a critical role in obesity development, the present study investigated the effect of an Hsp90 inhibitor radicicol on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service