Skip to Content
Merck
All Photos(1)

Documents

SML1147

Sigma-Aldrich

IQ-1

≥95% (HPLC)

Synonym(s):

2-[2-(4-Acetylphenyl)diazenyl]-2-(3,4-dihydro-3,3-dimethyl-1(2H)-isoquinolinylidene)-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N4O2
CAS Number:
Molecular Weight:
362.42
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(C1=CC=C(N=N/C(C(N)=O)=C2NC(C)(C)CC3=C/2C=CC=C3)C=C1)=O

InChI

1S/C21H22N4O2/c1-13(26)14-8-10-16(11-9-14)24-25-19(20(22)27)18-17-7-5-4-6-15(17)12-21(2,3)23-18/h4-11,23H,12H2,1-3H3,(H2,22,27)/b19-18+,25-24?

InChI key

ZXSRRUROLCGTBS-ASEVQSCHSA-N

Biochem/physiol Actions

IQ-1 enhances β-catenin/CBP interactions and blocks β-catenin/p300 driven transcription, which enables mouse embryonic stem cells (ESCs) to be maintained in an undifferentiated pluripotent state, allowing expansion and self-renewal of mouse ESC populations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service