Skip to Content
Merck
All Photos(1)

Key Documents

SML0350

Sigma-Aldrich

AZD-7762 hydrochloride

≥98% (HPLC)

Synonym(s):

5-(3-Fluorophenyl)-3-ureidothiophene-2-carboxylic acid N-[(S)-piperidin-3-yl]amide hydrochloride, AZD 7762 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19FN4O2S · HCl
CAS Number:
Molecular Weight:
398.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

Cl.NC(=O)Nc1cc(sc1C(=O)N[C@H]2CCCNC2)-c3cccc(F)c3

InChI

1S/C17H19FN4O2S.ClH/c18-11-4-1-3-10(7-11)14-8-13(22-17(19)24)15(25-14)16(23)21-12-5-2-6-20-9-12;/h1,3-4,7-8,12,20H,2,5-6,9H2,(H,21,23)(H3,19,22,24);1H/t12-;/m0./s1

InChI key

WFZBLOIXZRZEDG-YDALLXLXSA-N

Application

AZD-7762 hydrochloride has been used:
  • as a checkpoint kinase 1 and 2 (CHK1/2) inhibitor to study its effect on the efficiency of non-homologous end-joining (NHEJ) repair in breast cancer cell lines
  • as a CHK1 inhibitor to serve as a positive control for replication stress in the assessment of fork restart events
  • as a CHK1/2 inhibitor to study its effects on DNA lesions during mice zygote development

Biochem/physiol Actions

AZD-7762 is a potent, ATP-competitive inhibitor of Chk1 and Chk2 that enhanced cytotoxicity DNA-damaging agents.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Tonzi et al.
eLife, 7 (2018-11-14)
DNA replication stress is often defined by the slowing or stalling of replication fork progression leading to local or global DNA synthesis inhibition. Failure to resolve replication stress in a timely manner contribute toward cell cycle defects, genome instability and
Sabrina Ladstätter et al.
Cell, 167(7), 1774-1787 (2016-12-06)
Sexual reproduction culminates in a totipotent zygote with the potential to produce a whole organism. Sperm chromatin reorganization and epigenetic reprogramming that alter DNA and histone modifications generate a totipotent embryo. Active DNA demethylation of the paternal genome has been
Andrea M Pesch et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(24), 6568-6580 (2020-09-25)
Cyclin-dependent kinase 4/6 (CDK4/6) inhibitors have improved progression-free survival for metastatic, estrogen receptor-positive (ER+) breast cancers, but their role in the nonmetastatic setting remains unclear. We sought to understand the effects of CDK4/6 inhibition (CDK4/6i) and radiotherapy in multiple preclinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service