Skip to Content
Merck
All Photos(1)

Key Documents

SML1068

Sigma-Aldrich

TIC10 angular

≥98% (HPLC)

Synonym(s):

7-Benzyl-4-(2-methylbenzyl)-1,2,6,7,8,9-hexahydroimidazo[1,2-a]pyrido[3,4-e]pyrimidin-5(4H)-one., ONC-201, ONC201, TIC10 corrected

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N4O
Molecular Weight:
386.49
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

2-8°C

General description

TIC10 (TRAIL-inducing compound 10) is an anticancer molecule also referred as ONC201.

Biochem/physiol Actions

TIC10 (TRAIL-inducing compound 10) induces apoptosis mediated by TRAIL (TNF-related apoptosis-inducing ligand) in a number of cancers without toxicity such as breast, colon and glioblastoma multiforme tumors.
TIC10 angular is a potent blood brain barrier penetrant and orally active inducer of TRAIL production that induces death of multiple human cancers.
TIC10 angular is a potent blood brain barrier penetrant and orally active inducer of TRAIL production that induces death of multiple human cancers. TIC10 angular inactivates both Akt and ERK, which results in Foxo3a nuclear factor migration to the nucleus and binding to TRAIL promoter.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Small molecule ONC201/TIC10 targets chemotherapy-resistant colorectal cancer stem-like cells in an Akt/Foxo3a/TRAIL-dependent manner.
Varun V P, et al.
Cancer Research, 75(7), 1423-1432 (2015)
Jeremy R E
NMR in Pharmaceutical Science, 185-185 (2015)

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service