Skip to Content
Merck
All Photos(1)

Key Documents

SML0750

Sigma-Aldrich

TPPU

≥98% (HPLC)

Synonym(s):

1-Trifluoromethoxyphenyl-3-(1-propionylpiperidin-4-yl) urea, N-[1-(1-Oxopropyl)-4-piperidinyl]-N′-[4-(trifluoromethoxy)phenyl]-urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20F3N3O3
CAS Number:
Molecular Weight:
359.34
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

FC(F)(F)Oc1ccc(cc1)NC(=O)NC2CCN(CC2)C(=O)CC

InChI

1S/C16H20F3N3O3/c1-2-14(23)22-9-7-12(8-10-22)21-15(24)20-11-3-5-13(6-4-11)25-16(17,18)19/h3-6,12H,2,7-10H2,1H3,(H2,20,21,24)

InChI key

AAJMQTLFRTZCJK-UHFFFAOYSA-N

Application

TPPU has been used as a soluble epoxide hydrolase (SEH) inhibitor to characterize its pharmacokinetic (PK) and pharmacodynamic properties in rodents. It has also been used as a SEH inhibitor to study its effects on 12,13-dihydroxy-9Z-octadecenoic acid (12,13-DiHOME) concentrations in nervous tissues.

Biochem/physiol Actions

TPPU is a potent sEH inhibitor (IC50 = 3.7 nM) that has been shown to have very favorable PK attributes in cynomolgus monkeys. sEH converts epoxyeicosatrienoic acids (EETs) to dihydroxyiecosatrienoic acids (DHETs), and sEH inhibitors display anti-inflammatory and anti-atherosclerotic effects.
TPPU is a potent sEH inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annika I Ostermann et al.
Prostaglandins & other lipid mediators, 121(Pt A), 131-137 (2015-06-29)
Epoxides from polyunsaturated fatty acids (PUFAs) are potent lipid mediators. In vivo stabilization of these epoxides by blockade of the soluble epoxide hydrolase (sEH) leads to anti-inflammatory, analgesic and normotensive effects. Therefore, sEH inhibitors (sEHi) are a promising new class
The oxidized linoleic acid metabolite 12, 13-DiHOME mediates thermal hyperalgesia during inflammatory pain
Zimmer B, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1863(7), 669-678 (2018)
Oral treatment of rodents with soluble epoxide hydrolase inhibitor 1-(1-propanoylpiperidin-4-yl)-3-[4-(trifluoromethoxy) phenyl] urea (TPPU): Resulting drug levels and modulation of oxylipin pattern
Ostermann A I, et al.
Prostaglandins & other lipid mediators, 121 (2015)
Béla Zimmer et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1863(7), 669-678 (2018-04-07)
Eicosanoids play a crucial role in inflammatory pain. However, there is very little knowledge about the contribution of oxidized linoleic acid metabolites in inflammatory pain and peripheral sensitization. Here, we identify 12,13-dihydroxy-9Z-octadecenoic acid (12,13-DiHOME), a cytochrome P450-derived linoleic acid metabolite
Qiong Wu et al.
Journal of molecular neuroscience : MN, 67(3), 364-372 (2019-01-16)
High level of corticosterone (CORT) is toxic to neurons and plays an important role in depression-like behavior and chronic stress. Our previous study showed that TPPU, a soluble epoxide hydrolase (sEH) inhibitor (sEHI), induces an antidepressant effect in animal models.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service