Skip to Content
Merck
All Photos(1)

Key Documents

SML0517

Sigma-Aldrich

Amlexanox

≥98% (HPLC)

Synonym(s):

2-Amino-7-(1-methylethyl)-5-oxo-5H-[1]Benzopyrano[2,3-b]pyridine-3-carboxylic acid, AA 673, Amoxanox, CHX 3673

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O4
CAS Number:
Molecular Weight:
298.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL at warmed, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)c1ccc2Oc3nc(N)c(cc3C(=O)c2c1)C(O)=O

InChI

1S/C16H14N2O4/c1-7(2)8-3-4-12-9(5-8)13(19)10-6-11(16(20)21)14(17)18-15(10)22-12/h3-7H,1-2H3,(H2,17,18)(H,20,21)

InChI key

SGRYPYWGNKJSDL-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Amlexanox is an anti-allergic drug with anti-inflammatory properties.

Biochem/physiol Actions

Amlexanox elevates the amount of nonsense-containing mRNAs in treated cells and helps to generate full-length proteins effectively.
Amlexanox is an anti-allergy and anti-asthma drug that blocks histamine and leukotriene release from leukocytes and mast cells. Amlexanox has also been shown to inhibit cahaperone activity of Hsp90, and S100A13, which is involved in transport of proteins devoid of signal peptide sequences.
Amlexanox is an anti-allergy, anti-ulcer drug; S100A13 inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shannon M Reilly et al.
Nature medicine, 19(3), 313-321 (2013-02-12)
Emerging evidence suggests that inflammation provides a link between obesity and insulin resistance. The noncanonical IκB kinases IKK-ɛ and TANK-binding kinase 1 (TBK1) are induced in liver and fat by NF-κB activation upon high-fat diet feeding and in turn initiate
Miki Okada et al.
Biochemical and biophysical research communications, 292(4), 1023-1030 (2002-04-12)
S100 proteins are a multigenic family of low-molecular-weight Ca(2+)-binding proteins comprising 19 members. These proteins undergo a conformational change by Ca(2+)-binding and consequently interact with their target proteins. Recently, we reported that two antiallergic drugs, Amlexanox and Cromolyn, bind to
R W Barrons
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists, 58(1), 41-50 (2001-02-24)
The clinical features, etiology, and treatment of recurrent aphthous ulcers (RAU) are discussed. Aphthous ulcers are among the most common oral lesions in the general population, with a frequency of up to 25% and three-month recurrence rates as high as
M Sugiura et al.
Contact dermatitis, 38(2), 65-67 (1998-03-20)
A 23-year-old woman developed allergic rhinitis in 1993. Her doctor prescribed for her Solfa (amlexanox 25 mg) and Lysozymen 90 (lysozyme chloride 90 mg) tablets. She took the tablets occasionally, whenever she developed allergic rhinitis. She noticed a macular erythema
F Tarantini et al.
The Journal of biological chemistry, 276(7), 5147-5151 (2000-11-23)
Interleukin (IL)1alpha mediates proinflammatory events through its extracellular interaction with the IL1 type I receptor. However, IL1alpha does not contain a conventional signal peptide sequence that provides access to the endoplasmic reticulum-Golgi apparatus for secretion. Thus, we have studied the

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service