Skip to Content
Merck
All Photos(3)

Key Documents

377430

Sigma-Aldrich

Allyl isothiocyanate

95%

Synonym(s):

AITC, Oil of mustard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2NCS
CAS Number:
Molecular Weight:
99.15
Beilstein:
773748
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

contains

BHT as stabilizer

refractive index

n20/D 1.529 (lit.)

bp

150 °C (lit.)

mp

−80 °C (lit.)

density

1.013 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CCN=C=S

InChI

1S/C4H5NS/c1-2-3-5-4-6/h2H,1,3H2

InChI key

ZOJBYZNEUISWFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allyl isothiocyanate (AITC) was suitable for use in the following studies:
  • Quantitation of the N-acetylcysteine conjugate of AITC in human urine after ingestion of mustard.
  • Evaluation of the fungitoxic activity of AITC vapour on Penicillium expansum on pears, particularly on a thiabendazole (TBZ)-resistant P. expansum strain in vivo, to verify whether AITC treatment could be a substitute for TBZ.
  • Study of the induction of acetylation of histone in mouse erythroleukemia cells by some organosulfur compounds.

Biochem/physiol Actions

Naturally occurring constituent of cruciferous vegetables.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jill M Recla et al.
Pain, 160(8), 1740-1753 (2019-07-25)
Identification of genetic variants that influence susceptibility to pain is key to identifying molecular mechanisms and targets for effective and safe therapeutic alternatives to opioids. To identify genes and variants associated with persistent pain, we measured late-phase response to formalin
I M Bruggeman et al.
Toxicology and applied pharmacology, 83(2), 349-359 (1986-04-01)
Ally isothiocyanate has been reported to be a bladder carcinogen in male rats. On the other hand, benzyl isothiocyanate is an anti-carcinogen. These contradicting properties led us to investigate the cytotoxicity of these compounds in RL-4 rat hepatocytes. Since conjugation
Antifungal vapour-phase activity of allyl-isothiocyanate against Penicillium expansum on pears.
Mari M, et al.
Plant Pathology, 51(2), 231-236 (2002)
Chang Liu et al.
Neuron, 109(2), 273-284 (2020-11-06)
The TRPA1 ion channel is activated by electrophilic compounds through the covalent modification of intracellular cysteine residues. How non-covalent agonists activate the channel and whether covalent and non-covalent agonists elicit the same physiological responses are not understood. Here, we report
Ying Li et al.
Journal of colloid and interface science, 438, 130-137 (2014-12-03)
Allyl isothiocyanate (AITC), an organosulfur compound in cruciferous vegetables, is a natural antimicrobial and potential chemopreventive agent. However, the instability of AITC in aqueous systems restrains its applications. In this study, oil-in-water AITC nanoemulsion was prepared by the emulsion inversion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service