Skip to Content
Merck
All Photos(1)

Key Documents

SML0503

Sigma-Aldrich

9(S)-HODE

≥98% (HPLC)

Synonym(s):

9S-Hydroxy-10E,12Z-octadecadienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O3
CAS Number:
Molecular Weight:
296.44
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solution

concentration

1 mg/mL in ethanol

color

colorless to light yellow

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCC/C=C\C=C\[C@@H](O)CCCCCCCC(O)=O

InChI

1S/C18H32O3/c1-2-3-4-5-6-8-11-14-17(19)15-12-9-7-10-13-16-18(20)21/h6,8,11,14,17,19H,2-5,7,9-10,12-13,15-16H2,1H3,(H,20,21)/b8-6-,14-11+/t17-/m1/s1

InChI key

NPDSHTNEKLQQIJ-UINYOVNOSA-N

Biochem/physiol Actions

9(S)-HODE is produced by the lipoxygenation of linoleic acid. It is a major component of plasma oxidized low-density lipoprotein. 9-HODE has been shown to stimulate cell proliferation and and extracellular matrix synthesis in human mesangial cells via PPARgamma. Pro-inflammatory effects of 9-HODE, acting through the receptor GPR132, can contribute to atherosclerosis progression. 9-HODE has also been shown to activate spinal TRPV1, leading to inflammatory hyperalgesia.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shasha Li et al.
Scientific reports, 8(1), 14627-14627 (2018-10-04)
Zhenwu decoction (ZWD) is a promising traditional Chinese prescription against renal fibrosis, while its underlying mechanism remains unclear. Rat model of renal fibrosis were established and divided into control group, model group, ZWD treatment group and enalapril maleate treatment group.
[Activation mechanism of PPARgamma by its endogenous ligands].
Takuma Shiraki
Seikagaku. The Journal of Japanese Biochemical Society, 79(10), 960-964 (2007-11-22)
Holger Hübke et al.
Journal of agricultural and food chemistry, 53(5), 1556-1562 (2005-03-03)
The analysis of (R)-9- and (S)-9-hydroxy-10E,12Z-octadecadienoic acid as well as (R)-13- and (S)-13-hydroxy-9Z,11E-octadecadienoic acid (HODE) as free acids, esterified in triacylglycerols (storage lipids), and esterified in polar lipids (phospholipids, glycolipids, etc.) in barley, germinating barley, and finished malt was performed
Markus K Muellner et al.
The Biochemical journal, 420(2), 277-281 (2009-03-07)
LOOHs (lipid hydroperoxides) in oxLDL [oxidized LDL (low-density lipoprotein)] are potentially atherogenic compounds. Recently, H2S was identified as the third endogenous gasotransmitter in the vasculature. H2O2 is known to be destroyed by H2S. Assuming that H2S may also react with
Frédéric Coutant et al.
Journal of immunology (Baltimore, Md. : 1950), 172(1), 54-60 (2003-12-23)
Because of its oxidative modification during the acute-phase response to an aggression, low density lipoprotein (LDL) can be regarded as a source of lipid mediators that can act both to promote and inhibit inflammation. This can be exemplified by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service