Skip to Content
Merck
All Photos(1)

Key Documents

493732

Sigma-Aldrich

1,4-Butanediol

1,4-Butanediol

ReagentPlus®, 99%

Synonym(s):

1,4-Butylene glycol, Tetramethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)4OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1633445
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
230 °C (lit.)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor density

3.1 (vs air)

product line

ReagentPlus®

Assay

99%

form

solid or viscous liquid

autoignition temp.

698 °F

dilution

(for general lab use)

refractive index

n20/D 1.445 (lit.)

bp

230 °C (lit.)

mp

16 °C (lit.)

density

1.017 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCCCCO

InChI

1S/C4H10O2/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

WERYXYBDKMZEQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1,4-Butanediol can be used as a reactant to prepare:
  • Homoallyl alcohol (3-buten-1-ol) and tetrahydrofuran via vapor-phase dehydration using ZrO2 as a catalyst.[1]
  • Poly(1,4-butanediol succinate) polymer by reacting with succinic anhydride.[2]
  • Biodegradable homopolyesters [poly(butylene succinate) and poly(butylene adipate)] and copolyesters [poly(butylene succinate-co-butylene adipate)] by treating with succinic acid and adipic acid via esterification and deglycolization reaction.[3]
  • Poly(1,4-butyl sebacate) by polyesterification reaction with sebacic acid or its derivatives.[4]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

273.2 °F - closed cup

Flash Point(C)

134 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipase-catalyzed synthesis of poly (1, 4-butyl sebacate) from sebacic acid or its derivatives with 1, 4-butanediol
Linko Y-Y, et al.
Journal of Biotechnology, 40(2), 133-138 (1995)
Synthesis of homoallyl alcohol from 1, 4-butanediol over ZrO2 catalyst
Yamamoto N, et al.
Catalysis Communications, 6(7), 480-484 (2005)
Synthesis and characterization of biodegradable poly (1, 4-butanediol succinate)
Song DK and Sung YK
Journal of Applied Polymer Science, 56(11), 1381-1395 (1995)
Synthesis and characterization of the biodegradable copolymers from succinic acid and adipic acid with 1, 4-butanediol
Ahn BD, et al.
Journal of Applied Polymer Science, 82(11), 2808-2826 (2001)
Daehun Park et al.
Nature communications, 12(1), 263-263 (2021-01-13)
Clusters of tightly packed synaptic vesicles (SVs) are a defining feature of nerve terminals. While SVs are mobile within the clusters, the clusters have no boundaries consistent with a liquid phase. We previously found that purified synapsin, a peripheral SV

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service