Skip to Content
Merck
All Photos(1)

Documents

SML0450

Sigma-Aldrich

5-BDBD

≥98% (HPLC)

Synonym(s):

5-(3-Bromophenyl)-1,3-dihydro-2H-Benzofuro[3,2-e]-1,4-diazepin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11BrN2O2
CAS Number:
Molecular Weight:
355.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Brc1cccc(c1)C2=NCC(=O)Nc3c2oc4ccccc34

InChI

1S/C17H11BrN2O2/c18-11-5-3-4-10(8-11)15-17-16(20-14(21)9-19-15)12-6-1-2-7-13(12)22-17/h1-8H,9H2,(H,20,21)

InChI key

NKYMVQPXXTZHSF-UHFFFAOYSA-N

Biochem/physiol Actions

5-BDBD is a specific inhibitor of P2X4. The compound inhibits P2X4 currents in CHO cells with an IC50 of 500 nM.
In native tissues, 5-BDBD is considered as a beneficial antagonist to analyze the effect of P2X receptors (P2XRs) subtype P2X4R.

Features and Benefits

This compound is featured on the P2 Receptors: P2X Ion Channel Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Long et al.
Journal of neuroinflammation, 15(1), 245-245 (2018-09-01)
The mechanism underlying migraine chronification remains unclear. Central sensitization may account for this progression. The microglia P2X4 receptor (P2X4R) plays a pivotal role in the central sensitization of inflammatory and neuropathic pain, but there is no information about P2X4R in
Hyuck Joon Kwon et al.
Scientific reports, 6, 39302-39302 (2016-12-23)
Electrical stimulation (ES) is known to guide the development and regeneration of many tissues. However, although preclinical and clinical studies have demonstrated superior effects of ES on cartilage repair, the effects of ES on chondrogenesis remain elusive. Since mesenchyme stem
Bing Hu et al.
Iranian journal of basic medical sciences, 21(4), 364-369 (2018-05-26)
P2X4R is expressed in immunocyte and lung tissues. It has been a focus in inflammatory responses recently. This study investigated whether blockage of P2X4R attenuates allergic inflammation by modulating T cell response in ovalbumin-sensitized mice. Ovalbumin was used to sensitize
Claudio Coddou et al.
Neuroscience letters, 690, 219-224 (2018-10-27)
P2X receptors (P2XRs) are a family of ATP-gated ionic channels that are expressed in numerous excitable and non-excitable cells. Despite the great advance on the structure and function of these receptors in the last decades, there is still lack of
Yun-Ming Tu et al.
Food & function, 8(12), 4336-4346 (2017-09-25)
Circulating levels of free fatty acids (FFAs) are often found to be increased in patients with type 2 diabetes mellitus (T2DM) and metabolic syndrome (MS). High plasma FFA levels may give rise to maladaptive macrophage activation and promote inflammatory responses

Articles

We offer many products related to P2X ion channel family receptors for your research needs.

We offer many products related to P2X ion channel family receptors for your research needs.

We offer many products related to P2X ion channel family receptors for your research needs.

We offer many products related to P2X ion channel family receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service