Skip to Content
Merck
All Photos(1)

Documents

I8000010

Ivermectin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

22,23-Dihydroavermectin B1, MK-933

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ivermectin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]1(O[C@]2(CC[C@@H]1C)C[C@@H]3C[C@@H](C\C=C(C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C6/CO[C@]7([H])[C@H](O)C(C)=C[C@@H](C(=O)O3)[C@]67O)O2)[C@@H](C)CC

InChI

1S/C48H74O14/c1-11-25(2)43-28(5)17-18-47(62-43)23-34-20-33(61-47)16-15-27(4)42(26(3)13-12-14-32-24-55-45-40(49)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-10)44(31(8)57-39)60-38-21-36(53-9)41(50)30(7)56-38/h12-15,19,25-26,28,30-31,33-45,49-50,52H,11,16-18,20-24H2,1-10H3/b13-12+,27-15+,32-14+/t25-,26-,28-,30-,31-,33+,34-,35-,36-,37-,38-,39-,40+,41-,42-,43+,44-,45+,47+,48+/m0/s1

InChI key

AZSNMRSAGSSBNP-XPNPUAGNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ivermectin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ivermectin for scabies and headlice: veterinary use only.
Mark N Lowenthal
The Israel Medical Association journal : IMAJ, 15(5), 262-262 (2013-07-12)
Recurrent angioedema associated with secondary eosinophilia.
M Cugno et al.
Journal of investigational allergology & clinical immunology, 23(2), 129-130 (2013-05-10)
MMWR. Morbidity and mortality weekly report, 62(14), 264-266 (2013-04-12)
Strongyloides stercoralis is an intestinal nematode endemic in the tropics and subtropics. Immunocompetent hosts typically are asymptomatic, despite chronic Strongyloides infection. In contrast, immunocompromised patients are at risk for hyperinfection syndrome and disseminated disease, with a fatality rate >50%. The
Sébastien D S Pion et al.
PLoS neglected tropical diseases, 7(2), e2084-e2084 (2013-03-08)
Ivermectin has been the keystone of onchocerciasis control for the last 25 years. Sub-optimal responses to the drug have been reported in Ghanaian communities under long-term treatment. We assessed, in two Cameroonian foci, whether the microfilaricidal and/or embryostatic effects of
Postcard presentation: a dog with pruritus.
Compendium (Yardley, PA), 34(5), E7-E7 (2013-05-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service