Skip to Content
Merck
All Photos(1)

Key Documents

SML0416

Sigma-Aldrich

TCN-201

≥98% (HPLC)

Synonym(s):

3-Chloro-4-fluoro-N-[(4-{[2-(phenylcarbonyl)hydrazino]carbonyl}phenyl)methyl]benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17ClFN3O4S
CAS Number:
Molecular Weight:
461.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1Cl)S(=O)(=O)NCc2ccc(cc2)C(=O)NNC(=O)c3ccccc3

InChI

1S/C21H17ClFN3O4S/c22-18-12-17(10-11-19(18)23)31(29,30)24-13-14-6-8-16(9-7-14)21(28)26-25-20(27)15-4-2-1-3-5-15/h1-12,24H,13H2,(H,25,27)(H,26,28)

InChI key

FYIBXBFDXNPBSF-UHFFFAOYSA-N

Biochem/physiol Actions

TCN-201 is a sulphonamide derivative, that negatively modulates glycine binding to the GluN1 (glutamate receptor subunit) subunit of the receptor.
TCN-201 is the first NMDA receptor antagonist that is specific for NR2A-subunit containing receptors. The pIC50 for NR2A NMDA receptors is 6.8. TCN-201has no effect on NR2B-containing receptors at concentrations up to 50 mM.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural basis for negative allosteric modulation of GluN2A-containing NMDA receptors
Yi F, et al.
Neuron, 91(6), 1316-1329 (2016)
TCN 201 selectively blocks GluN2A-containing NMDARs in a GluN1 co-agonist dependent but non-competitive manner
Edman S, et al.
Neuropharmacology, 63(3), 441-449 (2012)
Jean-Baptiste Esmenjaud et al.
The EMBO journal, 38(2) (2018-11-07)
NMDA receptors (NMDARs) are glutamate-gated ion channels that are key mediators of excitatory neurotransmission and synaptic plasticity throughout the central nervous system. They form massive heterotetrameric complexes endowed with unique allosteric capacity provided by eight extracellular clamshell-like domains arranged as
Yan-Hai Li et al.
Neuroscience letters, 715, 134674-134674 (2019-12-07)
Medial vestibular nucleus (MVN) neurons are involved in the regulation of eye movements to endure the stability of the image during head movement, and play a critical role in plasticity of the vestibulo-ocular reflex (VOR) during the juvenile period. We
Giacomo Sitzia et al.
Neuropharmacology, 174, 108136-108136 (2020-06-01)
In Parkinson's disease (PD) reduced levels of dopamine (DA) in the striatum lead to an abnormal circuit activity of the basal ganglia and an increased output through the substantia nigra pars reticulata (SNr) and the globus pallidus internal part. Synaptic

Articles

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service