Skip to Content
Merck
All Photos(1)

Documents

B3809

Sigma-Aldrich

Benzenesulfonyl hydrazide

98%

Synonym(s):

BSH, Benzenesulfonylhydrazine, Porofor BSH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2NHNH2
CAS Number:
Molecular Weight:
172.20
Beilstein:
640079
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

mp

101-103 °C (lit.)

SMILES string

NNS(=O)(=O)c1ccccc1

InChI

1S/C6H8N2O2S/c7-8-11(9,10)6-4-2-1-3-5-6/h1-5,8H,7H2

InChI key

VJRITMATACIYAF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. D

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Symanczik et al.
Frontiers in plant science, 8, 1263-1263 (2017-08-05)
Naranjilla (
D Aberdein et al.
New Zealand veterinary journal, 65(6), 327-331 (2017-08-18)
AIMS To determine the frequency of the FAS-ligand gene (FASLG) variant associated with feline autoimmune lymphoproliferative syndrome (FALPS) and the proportion of carriers of the variant in three British shorthair (BSH) breeding catteries in New Zealand. METHODS Buccal swabs were
Peng Mi et al.
Journal of controlled release : official journal of the Controlled Release Society, 254, 1-9 (2017-03-25)
Boron neutron capture therapy is a promising tumor treatment method, though its wide application has been limited due to the poor tumor selectivity and intracellular delivery of
Martin S Lawless et al.
The Journal of the Acoustical Society of America, 142(4), 2258-2258 (2017-11-03)
Selecting an appropriate listening test design for concert hall research depends on several factors, including listening test method and participant critical-listening experience. Although expert listeners afford more reliable data, their perceptions may not be broadly representative. The present paper contains
Evgenii Zavjalov et al.
International journal of radiation biology, 96(7), 868-878 (2020-04-28)
Purpose: To evaluate the efficacy of boron neutron capture therapy (BNCT) for a heterotopic U87 glioblastoma model in SCID mice using boron phenylalanine (BPA), sodium borocaptate (BSH) and liposomal BSH as boron compounds at a unique, accelerator-based neutron source.Materials and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service