Skip to Content
Merck
All Photos(1)

Key Documents

SML0209

Sigma-Aldrich

ANA-12

≥98% (HPLC)

Synonym(s):

N-[2-[[(Hexahydro-2-oxo-1H-azepin-3-yl)amino]carbonyl]phenyl]-benzo[b]thiophene-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21N3O3S
CAS Number:
Molecular Weight:
407.49
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥5 mg/mL at 60 °C

storage temp.

2-8°C

SMILES string

O=C1NCCCCC1NC(=O)c2ccccc2NC(=O)c3cc4ccccc4s3

InChI

1S/C22H21N3O3S/c26-20(25-17-10-5-6-12-23-21(17)27)15-8-2-3-9-16(15)24-22(28)19-13-14-7-1-4-11-18(14)29-19/h1-4,7-9,11,13,17H,5-6,10,12H2,(H,23,27)(H,24,28)(H,25,26)

InChI key

TUSCYCAIGRVBMD-UHFFFAOYSA-N

Application

ANA-12 was used to inhibit TrkB in primary mouse cortical neurons and primary rat hippocampal cultures.

Biochem/physiol Actions

The neurotrophin brain-derived neurotrophic factor (BDNF) and its receptor tropomyosin-related kinase B (TrkB) have emerged as key mediators in the pathophysiology of several mood disorders, including anxiety and depression. ANA-12 is a TrkB ligand that prevents activation of the receptor by BDNF with a high potency. It binds selectively to TrkB and inhibits processes downstream of TrkB without altering TrkA and TrkC functions. In adult mice model ANA-12 decreases TrkB activity in the brain without affecting neuronal survival. Mice dosed with ANA-12 show reduced anxiety- and depression related behaviors on a variety of tests predictive of anxiolytic and antidepressant properties in humans. ANA-12 may be a valuable tool for studying BDNF/TrkB signaling and may constitute a lead compound for developing the next generation of therapeutic agents for the treatment of mood disorders. There is also N-T19, which is less potent.
TrkB antagonist that possesses potent anxiolytic-like and antidepressant-like activities in mice

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth T Barfield et al.
Frontiers in behavioral neuroscience, 11, 237-237 (2017-12-23)
Early-life trauma can increase the risk for, and severity of, several psychiatric illnesses. These include drug use disorders, and some correlations appear to be stronger in women. Understanding the long-term consequences of developmental stressor or stress hormone exposure and possible
Agnieszka Pałucha-Poniewiera et al.
Behavioural pharmacology, 30(6), 471-477 (2019-02-07)
Ketamine has been shown to induce a rapid antidepressant effect on patients with depression. In many animal models, both rapid and sustained antidepressant activities were also found in response to an antagonist of group II metabotropic glutamate receptors, LY341495, and
Maxime Cazorla et al.
The Journal of clinical investigation, 121(5), 1846-1857 (2011-04-21)
The neurotrophin brain-derived neurotrophic factor (BDNF) and its receptor tropomyosin-related kinase B (TrkB) have emerged as key mediators in the pathophysiology of several mood disorders, including anxiety and depression. However, therapeutic compounds that interact with TrkB receptors have been difficult
A Montalbano et al.
Hippocampus, 23(10), 879-889 (2013-05-16)
Long-term potentiation (LTP) is accompanied by increased spine density and dimensions triggered by signaling cascades involving activation of the neurotrophin brain-derived neurotrophic factor (BDNF) and cytoskeleton remodeling. Chemically-induced long-term potentiation (c-LTP) is a widely used cellular model of plasticity, whose
Shi-Meng Zhou et al.
Neuroreport, 28(5), 259-267 (2017-02-28)
Cucurbitacin IIa (CuIIa) is the major active component of the Helmseya amabilis root and is known to have antiviral and anti-inflammatory effects. In this study, we examined the antidepressant-like effects of CuIIa in a mouse model of chronic unpredictable mild

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service