Skip to Content
Merck
All Photos(1)

Key Documents

SML0130

Sigma-Aldrich

Loxiglumide

≥97% (HPLC)

Synonym(s):

4-[(3,4-dichlorobenzoyl)amino]-5-[(3-methoxypropyl)pentylamino]-5-oxo-pentanoic acid, CR 1505

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30Cl2N2O5
CAS Number:
Molecular Weight:
461.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

CCCCCN(CCCOC)C(=O)C(CCC(O)=O)NC(=O)c1ccc(Cl)c(Cl)c1

InChI

1S/C21H30Cl2N2O5/c1-3-4-5-11-25(12-6-13-30-2)21(29)18(9-10-19(26)27)24-20(28)15-7-8-16(22)17(23)14-15/h7-8,14,18H,3-6,9-13H2,1-2H3,(H,24,28)(H,26,27)

InChI key

QNQZBKQEIFTHFZ-UHFFFAOYSA-N

Biochem/physiol Actions

Loxiglumide is a CCK-A receptor antagonist
Loxiglumide is a small-molecule antagonist of the cholecystokinin receptor CCKA. Loxiglumide inhibits pancreatic secretion of digestive enzymes, and also blocks CCK-induced gastric secretions and emptying.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salvatore Fanali et al.
Journal of chromatography. A, 1051(1-2), 247-252 (2004-11-10)
Hepta-Tyr antibiotic modified silica stationary phase was used for the chiral resolution of D,L-loxiglumide, a new drug under investigation proposed for the treatment of gastrointestinal diseases. The chiral stationary phase was packed into fused silica capillaries of 75 microm i.d.
Stefania De Luca et al.
Journal of medicinal chemistry, 49(8), 2456-2462 (2006-04-14)
The interaction between the 1-47 N-terminus of the CCK(1)-R and the anthranilic acid based antagonists has been investigated by fluorescence spectroscopy. These antagonists interact with W39 of the N-terminal domain of the CCK(1)-R like that of the endogenous ligand CCK-8.
D P Hirsch et al.
Digestive diseases and sciences, 47(11), 2531-2537 (2002-11-28)
To reduce weight, some morbidly obese patients are treated with an intragastric balloon, often resulting in increased reflux symptoms. As transient lower esophageal sphincter relaxations (TLESRs) are the major mechanism underlying reflux and can be reduced by cholecystokinin-A (CCK(A)) blockade
Shlomo Yehuda et al.
Nutritional neuroscience, 7(2), 85-90 (2004-07-29)
Iron deficiency (ID) is among the most common nutritional diseases, causing deleterious effects that include decreases in cognitive function and weight loss. The ID also induces a reduction in the number and affinity of dopaminergic D2 receptors. The new finding
Kaoru Ishizaki et al.
Pancreas, 26(1), 87-91 (2002-12-25)
To examine the involvement of cholecystokinin (CCK) in the basal pancreatic exocrine, we investigated the effect of loxiglumide (CR1505), a CCK1 receptor antagonist, on basal pancreatic exocrine secretion in conscious rats. After the basal collection of pancreatic juice for 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service