Skip to Content
Merck
All Photos(1)

Key Documents

SML3575

Sigma-Aldrich

TY-51469

≥98% (HPLC)

Synonym(s):

2-[4-[[(5-Fluoro-3-methylbenzo[b]thien-2-yl)sulfonyl]amino]-3-(methylsulfonyl)phenyl]-4-thiazolecarboxylic acid, TY 51469

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15FN2O6S4
CAS Number:
Molecular Weight:
526.60
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC2=C(C=C1)SC(S(NC3=CC=C(C=C3S(C)(=O)=O)C4=NC(C(O)=O)=CS4)(=O)=O)=C2C

Biochem/physiol Actions

TY-51469 is a potent and selective chymase inhibitor that prevents development and progression of non-alcoholic steatohepatitis in rats. TY-51469 inhibits activation of intestinal MMP-9 and prevents intestinal damage in indomethacin-induced small intestinal damage in rats. Also TY-51469 might protect against pancreatic islet disorganization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Significance of chymase-dependent matrix metalloproteinase-9 activation on indomethacin-induced small intestinal damages in rats
Kakimoto K, Takai S, Murano M, Ishida K, Yoda Y, Inoue T, Jin D, Umegaki E, Higuchi K
Journal of Pharmacology and Experimental Therapeutics, 332, 684-689 (2010)
Chymase inhibitor prevents the development and progression of non-alcoholic steatohepatitis in rats fed a high-fat and high-cholesterol diet
Miyaoka Y, Jin D, Tashiro K, Komeda K, Masubuchi S, Hirokawa F, Hayashi M, Takai S, Uchiyama K
Journal of Pharmacological Sciences, 134, 139-146 (2017)
Chymase inhibition reduces infarction and matrix metalloproteinase-9 activation and attenuates inflammation and fibrosis after acute myocardial ischemia/reperfusion
Oyamada S, Bianchi C, Takai S, Chu LM, Sellke FW
Journal of Pharmacology and Experimental Therapeutics, 339, 143-151 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service