Skip to Content
Merck
All Photos(1)

Key Documents

SBPE03

Sigma-Aldrich

SB-MRP2-Sf9-PREDEASY-ATPase kit

membrane based kit for Human MRP2 ATPase Transport Assays

Synonym(s):

ABCC2, MRP2, MRP2 human membrane based kit for Human MRP2 ATPase Transport Assays, Multidrug resistance protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

recombinant

expressed in insect cells

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ABCC2(1244)

General description

SOLVO Biotechnology offers ready-to-use assay kits for efflux and uptake transporters. SOLVO′s PREDEASY ATPase Assay Kit provides everything necessary to conduct ATPase activation and inhibition assays to better understand the nature of the interaction between a compound and the transporter. By measuring ATPase activity, both activation and inhibition of transporters can be investigated.

Application

SOLVO′s PREDEASY ATPase Assay Kit is a unique product for measuring the ATPase-dependent drug - ABC membrane transporter interaction in a convenient, "ready-to-assay" package. The ATPase assay is an in vitro membrane assay designed to indicate the nature of the interaction between the compound and the transporter. By measuring ATPase activity, both activation and inhibition of transporters can be investigated using membranes from baculovirus-infected insect cells or mammalian cell membranes containing high levels of human or rodent wild-type transporters. ABC transporters mediate the transport of substrates against a concentration gradient using energy derived from ATP hydrolysis, which is proportional to the transporter activity and could easily be detected with a colorimetric method.

PREDIVEZ PREDEASY Kits contain all reagents (chemicals) and solutions necessary to conduct ATPase activation and inhibition assays. You only have to add water. Each kit comes with a CD that includes the assay protocol, data sheet, SDS, electronic data processing, evaluation templates and the reference data. The recommended layout allows activation and inhibition measurements in duplicates for 8 concentrations of the test article on one 96-well plate with all of the relevant controls.

PREDEASY ATPase Assay Kits are available for 3 and 10 plates for the transporters recommended by the FDA and the EMA.

Physical form

Supplied as frozen membrane vesicles, containing 5 mg/ml membrane protein, labeled with volume, catalog number (transporter), date of production, protocols and necessary reagents sufficient for the analysis of 3 or 10, 96-well plates.

Legal Information

Distributed for SOLVO Biotechnology, Inc.
PREDEASY is a trademark of Solvo Biotechnology
PREDIVEZ is a trademark of Solvo Biotechnology

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Evers et al.
British journal of cancer, 83(3), 375-383 (2000-08-06)
The multidrug resistance proteins MRP1 and MRP2 are members of the same subfamily of ATP-binding cassette transporters. Besides organic molecules conjugated to negatively charged ligands, these proteins also transport cytotoxic drugs for which no negatively charged conjugates are known to
Thomas Rau et al.
Clinical pharmacology and therapeutics, 80(5), 468-476 (2006-11-23)
The adenosine triphosphate-binding cassette (ABC) class transporter ABCC2 (MRP2 [multidrug resistance related protein 2] or cMOAT [canalicular multispecific organic anion transporter]) is involved in the cellular outward transport and elimination of methotrexate. We hypothesized that common genetic variations may contribute
Potentiation of MRP2/Mrp2-mediated estradiol-17beta-glucuronide transport by drugs--a concise review.
Krisztina Herédi-Szabó et al.
Chemistry & biodiversity, 6(11), 1970-1974 (2009-11-26)
Isabelle Benz-de Bretagne et al.
Journal of biomedicine & biotechnology, 2011, 498757-498757 (2011-05-05)
MRP2 encoded by ABCC2 gene is involved in the secretion of numerous drugs and endogenous substrates. Patients with Dubin-Johnson syndrome due to mutation in ABCC2 gene have elevated urinary coproporphyrin ratio (UCP I/(I + III)). Here we investigated whether this
Anne T Nies et al.
Pflugers Archiv : European journal of physiology, 453(5), 643-659 (2006-07-19)
ABCC2 is a member of the multidrug resistance protein subfamily localized exclusively to the apical membrane domain of polarized cells, such as hepatocytes, renal proximal tubule epithelia, and intestinal epithelia. This localization supports the function of ABCC2 in the terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service