Merck
  • Vinblastine and sulfinpyrazone export by the multidrug resistance protein MRP2 is associated with glutathione export.
Vinblastine and sulfinpyrazone export by the multidrug resistance protein MRP2 is associated with glutathione export.
British journal of cancer (2000-08-06)
R Evers, M de Haas, R Sparidans, J Beijnen, P R Wielinga, J Lankelma, P Borst
ABSTRACT
The multidrug resistance proteins MRP1 and MRP2 are members of the same subfamily of ATP-binding cassette transporters. Besides organic molecules conjugated to negatively charged ligands, these proteins also transport cytotoxic drugs for which no negatively charged conjugates are known to exist. In polarized MDCKII cells, MRP1 routes to the lateral plasma membrane, and MRP2 to the apical plasma membrane. In these cells MRP1 transports daunorubicin, and MRP2 vinblastine; both transporters export reduced glutathione (GSH) into the medium. We demonstrate that glutathione transport in MDCKII-MRP1 cells is inhibited by the inhibitors of organic anion transporters sulfinpyrazone, indomethacin, probenecid and benzbromarone. In MDCKII-MRP2 cells, GSH export is stimulated by low concentrations of sulfinpyrazone or indomethacin, whereas export is inhibited down to control levels at high concentrations. We find that unmodified sulfinpyrazone is a substrate for MRP2, also at concentrations where GSH export is inhibited. We also show that GSH export in MDCKII-MRP2 cells increases in the presence of vinblastine, and that the stoichiometry between drug and GSH exported is between two and three. Our data indicate that transport of sulfinpyrazone and vinblastine is associated with GSH export. However, at high sulfinpyrazone concentrations this compound is transported without GSH. Models of MRP action are discussed that could explain these results.
Sigma-Aldrich
SB-ratMrp2-HEK293, MRP2 rat vesicles
Sigma-Aldrich
SB-MRP2-Sf9, membrane preparation for Vesicular Transport Assays, recombinant, expressed in baculovirus infected Sf9 cells
Sigma-Aldrich
SB-MRP2-Sf9-PREDEASY-ATPase kit, membrane based kit for Human MRP2 ATPase Transport Assays
Sigma-Aldrich
SB-MRP2-HEK293