Skip to Content
Merck
All Photos(1)

Key Documents

SAMVP64BSTV

Sigma-Aldrich

dCas9-VP64-Blasticidin SAM CRISPRa Helper Construct 1 Lentiviral Transduction Particles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

packaging

pkg of 8x25 μL (vials)

concentration

≥1x106 VP/ml (via p24 assay)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

General description

Ready to use dCas9-VP64 lentiviral particles enable immediate transduction of a wide range of cell lines. dCas9-VP64 lentiviral particles efficiently and stably integrate dCas9-VP64 and blasticidin resistance cassette linked by a 2A peptide and driven by the EF1 alpha promoter (EF1a-dCas9-VP64-2A-Blasticidin) for strong expression in both dividing and non-dividing cell lines. They are ideal for avoiding the tedious lentivirus production process and are one part of a three part CRISPR system with individual dCas9-VP64. MS2-p65-HSF1 and gRNA expression vectors.

To order gRNA in any format click here

Application

Functional Genomics/Target Validation
  • Unbiased forward genetic screening
  • Strong transcriptional activation in multiple cell lines
  • Creation of cell lines stably expressing dCas9-VP64.

Features and Benefits

  • Highly specific and highly active
  • Sequence verified high purity, high titer lentiviral particles
  • Activates genes through transcriptional activation rather than cDNA based overexpression

Learn more about SAM CRISPR Activators at SigmaAldrich.com/CRISPRa

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be rendered inactive (dCas9) with mutations to the two protein domains, RuvC and HnH (D10A and H840A respectively), which are responsible for nuclease activity. The nuclease deficient protein can then be fused with the transcriptional activator VP64 and used in conjunction with a guide RNA modified with MS2 RNA aptamers that function to recruit the additional transcriptional coactivators p65 and HSF1. The assembled SAM complex is then used as a cargo delivery system to target gene promoters, enabling site-specific transcriptional activation of the gene of interest.

Unit Definition

VP/mL is the concentration unit of measure for viral titer estimated by p24 assay.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service