Skip to Content
Merck
All Photos(1)

Key Documents

SAMHELPERV

Sigma-Aldrich

SAM CRISPRa Helper Construct Kit Lentiviral Transduction Particles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51
Pricing and availability is not currently available.

packaging

pkg of 16 vials (8x25μL aliquots for each of the 2 kit components)

concentration

≥1x106 VP/ml (via p24 assay)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

Related Categories

General description

This product is a kit containing two sets of ready to use lentiviral particles enabling immediate transduction for strong expression in a wide range of both dividing and non-dividing cell lines. MS2-P65-HSF1 lentiviral particles efficiently and stably integrate MS2-P65-HSF1 and hygromycin resistance cassette linked by a 2A peptide and driven by the EF1 alpha promoter (EF1a-dCas9-VP64-2A-Blasticidin). dCas9-VP64 lentiviral particles efficiently and stably integrate dCas9-VP64 and blasticidin resistance cassette linked by a 2A peptide and driven by the EF1 alpha promoter (EF1a-dCas9-VP64-2A-Blasticidin). They are ideal for avoiding the tedious lentivirus production process and are part of a three part CRISPR system with individual dCas9-VP64. MS2-p65-HSF1 and gRNA expression vectors.

To order gRNA in any format click here

Application

Functional Genomics/Target Validation
  • Unbiased forward genetic screening
  • Strong transcriptional activation in multiple cell lines
  • Creation of cell lines stably expressing dCas9-VP64 and MS2-p65-HSF1.

Features and Benefits

  • Highly specific and highly active
  • Sequence verified high purity, high titer lentiviral particles
  • Activates genes through transcriptional activation rather than cDNA based overexpression

Learn more about SAM CRISPR Activators at SigmaAldrich.com/CRISPRa

Components

This kit contains 2 components :
2 Helper Constructs with a minimum p24 titer of 1x106 VP/mL
  • 8 x 25μl vials of dCas9-VP64-Blasticidin SAM CRISPRa Helper Construct 1 Lentiviral Transduction Particles
  • 8 x 25μl vials of MS2-P65-HSF1-Hygromycin SAM CRISPRa Helper Construct 2 Lentiviral Transduction Particles

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be rendered inactive (dCas9) with mutations to the two protein domains, RuvC and HnH (D10A and H840A respectively), which are responsible for nuclease activity. The nuclease deficient protein can then be fused with the transcriptional activator VP64 and used in conjunction with a guide RNA modified with MS2 RNA aptamers that function to recruit the additional transcriptional coactivators p65 and HSF1. The assembled SAM complex is then used as a cargo delivery system to target gene promoters, enabling site-specific transcriptional activation of the gene of interest.[1]

Unit Definition

VP/mL is the concentration unit of measure for viral titer estimated by p24 assay.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-scale transcriptional activation by an engineered CRISPR-Cas9 complex
Konermann S, et al.
Nature, 517, 583-588 (2015)
Genome-scale CRISPR-Cas9 Knockout and Transcriptional Activation Screening.
Joung, S. et al.
Nature Protocols, 12, 828-863 (2017)
Julia Joung et al.
Nature protocols, 12(4), 828-863 (2017-03-24)
Forward genetic screens are powerful tools for the unbiased discovery and functional characterization of specific genetic elements associated with a phenotype of interest. Recently, the RNA-guided endonuclease Cas9 from the microbial CRISPR (clustered regularly interspaced short palindromic repeats) immune system

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service