Skip to Content
Merck
All Photos(1)

Documents

SAB5500082

Sigma-Aldrich

Anti-Chromogranin A antibody, Rabbit monoclonal

clone SP12, recombinant, expressed in proprietary host, tissue culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

SP12, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

bovine, pig

technique(s)

immunohistochemistry: 1:200

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CHGA(1113)

General description

Chromogranin A (CgA) is a circulating protein liberated by the neuroendocrine system. It is a glycoprotein that is located on human chromosome 14.
Chromogranin A (a protein of 439 amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, phenochromocytomas, paragangliomas, and other neuroendocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Immunogen

Recombinant protein encoding human chromogranin A.

Biochem/physiol Actions

Chromogranin A (CgA) controls vascular physiology and angiogenesis. It modulates the storage and liberation of catecholamine via intracellular (vesiculogenic) and extracellular (catecholamine release–inhibitory) mechanisms. CHGA is essential to generate catecholamine secretory vesicles in chromaffin-lineage PC12 cells and is required to promote a regulated secretion system in nonsecretory cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody supplied as tissue culture supernatant in TBS/1% BSA buffer pH 7.5 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasma levels of vasostatin-1, a chromogranin A fragment, are associated with carotid artery maximum stenosis: A pilot study.
Bachetti T, et al.
International Journal of Cardiology, 236, 438-443 (2017)
Both rare and common polymorphisms contribute functional variation at CHGA, a regulator of catecholamine physiology.
Wen G, et al.
American Journal of Human Genetics, 74(2), 197-207 (2004)
The human chromogranin A gene: chromosome assignment and RFLP analysis.
Modi WS, et al.
American Journal of Human Genetics, 45(5), 814-814 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service