Skip to Content
Merck
All Photos(3)

Documents

SAB4502906

Sigma-Aldrich

Anti-Synaptophysin antibody produced in rabbit

affinity isolated antibody

Synonym(s):

major synaptic vesicle protein p38, synaptophysin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 33 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYP(6855)

General description

Synaptophysin (SYP) is an integral membrane protein of synaptic vesicles. It is expressed in cortical synaptic terminals, during neurogenesis. The gene is located on human chromosome Xp11.23.
Synaptophysin is an integral membrane glycoprotein that is localized in presynaptic vesicles of nerve cells. This protein may regulate exocytosis and the formation of synaptic vesicles . Synaptophysin is encoded by the SYP gene and genetic alterations in SYP can lead to mental retardation . Anti-synaptophysin antibody specifically detects endogenous levels of total synaptophysin protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Synaptophysin.

Immunogen Range: 101-150

Application

Anti-Synaptophysin antibody produced in rabbit has been used in:
  • immunocytochemistry
  • western blotting
  • immunohistochemistry
Anti-synaptophysin antibody is suitable for use in immunohistochemistry and western blot.

Biochem/physiol Actions

Synaptophysin (SYP) modulates neurotransmitter release and synaptic plasticity. It maintains biogenesis and recycling of synaptic vesicles. SYP functions as a marker for neuroendocrine tumors. It is used as a potent marker for neuroendocrine prostate cancer (NEPC). Mutations in this gene is associated with X-linked intellectual disability.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single cell transcriptome analysis of mouse carotid body glomus cells
Zhou T, et al.
The Journal of Physiology, 594(15), 4225-4251 (2016)
Yaoming Wang et al.
Nature medicine, 22(9), 1050-1055 (2016-08-23)
Activated protein C (APC) is a blood protease with anticoagulant activity and cell-signaling activities mediated by the activation of protease-activated receptor 1 (F2R, also known as PAR1) and F2RL1 (also known as PAR3) via noncanonical cleavage. Recombinant variants of APC
Synaptophysin expression on circulating tumor cells in patients with castration resistant prostate cancer undergoing treatment with abiraterone acetate or enzalutamide
Pal SK, et al.
Urologic Oncology, 36(4), 162-e1-162-e1 (2018)
Moderate maternal food restriction in mice impairs physical growth, behavior, and neurodevelopment of offspring
Akitake Y, et al.
Nutrition Research (New York, N.Y.), 35(1), 76-87 (2015)
Lei Zhang et al.
eNeuro, 4(6) (2017-12-09)
The dendritic tree is a key determinant of neuronal information processing. In the motor system, the dendritic tree of spinal cord neurons undergoes dramatic remodeling in an activity-dependent manner during early postnatal life. This leads to the proper segmental spinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service