Skip to Content
Merck
All Photos(1)

Documents

SAB4700170

Sigma-Aldrich

Monoclonal Anti-CD38 antibody produced in mouse

clone HIT2, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HIT2, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD38(952)

General description

The antibody HIT2 reacts with CD38 (T10), a 45 kDa type II transmembrane glycoprotein strongly expressed mainly on plasma cells and activated T and B lymphocytes; it is an antigenic marker of lymphoid cells.

Immunogen

Human thymocytes in foetus

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 2-8 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Cluster of differentiation 38 (CD38) controls the release of oxytocin hormone that regulates social cognition and affiliative behaviors. CD38 serves as a receptor, an ectoenzyme and acts as a highly conserved calcium-signaling molecule while functioning as a secondary messenger. The generation of cyclic Adenosine diphosphate-ribose and nicotinic acid adenine dinucleotide phosphate from NAD+ and NAD phosphate by ADP-ribosyl cyclase is catalyzed by CD38. The formation of adenosine is an important function of CD38. The pleiotropic nucleoside adenosine formed, acts as a regulator of immune functions and inflammation process.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ADP ribosyl-cyclases (CD38/CD157), social skills and friendship.
Chong A, et al.
Psychoneuroendocrinology, 78, 185-192 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service