Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500072

Sigma-Aldrich

Anti-p16 INK antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD2A1, CDK4I, CDKN2, CDKN2A, CDN2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 16 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDKN2A(1029)

Related Categories

General description

Anti-p16 INK Antibody detects endogenous levels of total p16 INK protein.
Cyclin dependent kinase inhibitor 2A (CDKN2A) also known as P16-INK4A, belongs to the Ink4 (inhibitor of CDK4) family of CDK inhibitors. It is encoded by the gene mapped to human chromosome 9p21 within the INK4a / alternate reading frame (ARF) locus. Human P16-INK4A is localized in both nucleus and cytoplasm, and its expression increases with ageing in skin and kidney tissues.

Immunogen

The antiserum was produced against synthesized peptide derived from human p16 INK.

Immunogen Range: 10-59

Application

Anti-p16 INK, N-Terminal antibody produced in rabbit has been used in western blot analysis.

Biochem/physiol Actions

Cyclin dependent kinase inhibitor 2A (CDKN2A) also known as P16-INK4A, arrests cell cycle progression by inhibiting S-phase and promotes apoptosis. The encoded protein has anti-proliferative biological activity, and is implicated in the retinoblastoma (Rb) protein pathway. P16-INK4A acts as a tumor suppressor and plays a vital role in ageing and cellular senescence. Increased expression of this protein might lead to various types of high grade cancers. P16-INK4A also facilitates cell invasion and angiogenesis. Bronchial lavage P16INK4A promoter methylation might function as a potential biomarker for diagnosis of lung cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression and significance of p53, rb, p21/waf-1, p16/ink-4a, and PTEN tumor suppressors in canine melanoma.
Koenig A, et al.
Veterinary Pathology, 39(4), 458-472 (2002)
Antiproliferative effects of local anesthetics on mesenchymal stem cells: potential implications for tumor spreading and wound healing.
Lucchinetti E, et al.
Anesthesiology, 116(4), 841-856 (2012)
Ping Yang et al.
Journal of cellular and molecular medicine, 28(11), e18388-e18388 (2024-05-31)
Atherosclerosis, characterized by the accumulation of lipid plaques on the inner walls of arteries, is the leading cause of heart attack, stroke and severe ischemic injuries. Senescent cells have been found to accumulate within atherosclerotic lesions and contribute to the
p16(Ink4a) overexpression in cancer: a tumor suppressor gene associated with senescence and high-grade tumors.
Romagosa C, et al.
Oncogene, 30(18), 2087-2097 (2011)
Bronchial lavage P 16INK4A gene promoter methylation and lung cancer diagnosis: A meta-analysis.
Yifan D, et al.
Indian Journal of Cancer, e96-e98 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service