Skip to Content
Merck
All Photos(14)

Key Documents

ZRB1141

Sigma-Aldrich

Anti-p21 Antibody, clone 5G7, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative
Anti-p21 Antibody, clone 5G7, ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-CAP20, Anti-CDKN1, Anti-CIP1, Anti-MDA-6, Anti-P21, Anti-SDI1, Anti-WAF1, Anti-p21CIP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5G7, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 18.12 kDa
observed mol wt ~19 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CDKN1A(1026)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 5G7 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects p21/Waf1/Cip1 protein.

Immunogen

His-tagged full-length recombinant human Cyclin-dependent kinase inhibitor 1 (p21).

Application

Anti-p21, clone 5G7 ZooMAb , Cat. No. ZRB1141, is a recombinant rabbit monoclonal antibody that specifically targets p21-Waf1-Cip1 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected p21 in HUVEC and NIH 3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected p21 in human cervical cancer tissue sections.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

Cyclin-dependent kinase inhibitor 1 (UniProt: P38936; also known as CDK-interacting protein 1, Melanoma differentiation-associated protein 6, MDA-6, p21) is encoded by the CDKN1A (also known as CAP20, CDKN1, CIP1, MDA6, PIC1, SDI1, WAF1) gene (Gene ID:1026) in human. p21 belongs to the Cip and Kip family of CDK inhibitors. It binds to and inhibits cyclin-dependent kinase activity, preventing phosphorylation of critical cyclin-dependent kinase substrates and blocking cell cycle progression. It binds tightly to the G1 and S phase kinases, cyclin E/Cdk2, cyclin D/Cdk4, and cyclin A/Cdk2, and effectively inhibits their activity. However, its effects on G2/M kinases is minimal. It binds the cyclin subunit through a conserved Cy1 motif in the N-terminal half and through a weaker and Cy2 motif in the C-terminal half. p21 also inhibits proliferating cell nuclear antigen (PCNA) and binds to and blocks the inactivation of retinoblastoma protein (Rb), which is essential for cell cycle progression. It is an important effector of p53 and is important in the DNA-damage checkpoint. It is expressed in all adult tissues, with 5-fold lower levels observed in the brain. Is cyclin binding site is localized to amino acids 17-24 and CDK binding occurs in the region of 53-58 residues. p21 is shown to suppress tumor growth by promoting cell cycle arrest in response to various stimuli. However, it is often misregulated in various human cancers. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Abbas, T., and Dutta, A (2009). Nat Rev Cancer. 9(6); 400-414).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yogev Sela et al.
eLife, 10 (2021-02-24)
Cancer patients often harbor occult metastases, a potential source of relapse that is targetable only through systemic therapy. Studies of this occult fraction have been limited by a lack of tools with which to isolate discrete cells on spatial grounds.
Nan-Yu Zou et al.
Bone research, 12(1), 6-6 (2024-01-25)
Skeletal stem/progenitor cell (SSPC) senescence is a major cause of decreased bone regenerative potential with aging, but the causes of SSPC senescence remain unclear. In this study, we revealed that macrophages in calluses secrete prosenescent factors, including grancalcin (GCA), during
Yoshihiko Kobayashi et al.
Nature cell biology, 22(8), 934-946 (2020-07-15)
Stem cells undergo dynamic changes in response to injury to regenerate lost cells. However, the identity of transitional states and the mechanisms that drive their trajectories remain understudied. Using lung organoids, multiple in vivo repair models, single-cell transcriptomics and lineage
Daan M K van Soest et al.
Nature communications, 15(1), 2725-2725 (2024-03-29)
Reactive Oxygen Species (ROS) derived from mitochondrial respiration are frequently cited as a major source of chromosomal DNA mutations that contribute to cancer development and aging. However, experimental evidence showing that ROS released by mitochondria can directly damage nuclear DNA

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

Active Filters

  1. 神奈川
    • Review 1
    • Votes 0
    5 out of 5 stars.

    問題なく機能しました

    老齢マウス組織のIHC-P(希釈倍率: 1/200)およびHUVECでのウェスタンブロット(希釈倍率: 1/5,000)で良好に使用できました。

    Translate with Google

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service